Documentos clave
WHA10334500
Whatman® prepleated qualitative filter paper for technical use, Grade 0860 1/2
L × W (100 mm) (300 mm), pack of 500 ea
About This Item
Código UNSPSC:
41115727
NACRES:
NB.24
envase
pack of 500 ea
fabricante / nombre comercial
Cytiva
Whatman 10334500
Parámetros
60 sec/100 mL speed (Herzberg)
L × An
(100 mm) (300 mm)
grosor
170 μm
tamaño de poro
7-12 μm (Particle retention)
peso básico
75 g/m2
Descripción general
Time-saving Whatman qualitative grades are offered in folded (prepleated) form, which gives major advantages over flat circles:
  • Savings in time required to quadrant-fold circles to fit conical filter funnels in repetitive or multiple analyses
  • Decreased overall filtration time because of the extra surface area exposed; the normal slow down of filtration speed due to the loading of particulate is postponed
  • Increased total loading capacity as more filter area is available
  • Maintained flow rate due to the reduction in filter paper contact with funnel side and, the self-supporting shape of the filter itself
  • The prepleating does not significantly affect any of the technical data and the same figures may be used for the flat circles
Grade 0860 ½ : 7-12 μm (medium fast, smooth filter paper)
Comparable to grade 0858 but with a smooth surface, slightly thinner and faster.
Información legal
Whatman is a registered trademark of Cytiva
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)