Merck
R496065
N,N'-DI-PYRIDIN-2-YL-BENZENE-1,2-DIAMINE
AldrichCPR
Seleccione un Tamaño
10MG
US$ 35,60
US$ 35,60
PRECIOS SIN IMPUESTOS NACIONALES
Envío en 2 días laborables (Para pedidos fuera de Estados Unidos, calcule la entrega 1 o 2 semanas más tarde)
Seleccione un Tamaño
Cambiar Vistas
10MG
US$ 35,60
About This Item
Fórmula lineal:
C16H14N4
Número de CAS:
Peso molecular:
262.317
Número MDL:
US$ 35,60
PRECIOS SIN IMPUESTOS NACIONALES
Envío en 2 días laborables (Para pedidos fuera de Estados Unidos, calcule la entrega 1 o 2 semanas más tarde)
Otras notas
Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of rare and unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.
NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH'S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT AS-IS AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)