Documentos clave
DWK73802-13415
KIMBLE® solid cap with PTFE faced rubber liner
black phenolic cap, cap for 13-415
Seleccione un Tamaño
1 EACH
US$ 202,00
US$ 202,00
PRECIOS SIN IMPUESTOS NACIONALES
Comprobar disponibilidad del carrito
Seleccione un Tamaño
Cambiar Vistas
1 EACH
US$ 202,00
About This Item
Código UNSPSC:
41121800
NACRES:
NB.17
US$ 202,00
PRECIOS SIN IMPUESTOS NACIONALES
Comprobar disponibilidad del carrito
Materiales
PTFE/rubber liner
black phenolic cap
esterilidad
non-sterile
Características
class A
envase
case of 500 ea
fabricante / nombre comercial
Kimble® 73802-13415
Adaptador
cap for 13-415
Descripción general
Class A Caps with PTFE-Faced Rubber Liners highly resistant to temperature and steam during autoclaving
  • Specially formulated phenolic cap material
  • Autoclavabl
  • White rubber liners
  • Excellent for general laboratory use
  • Identified in ASTM Specification E982, Class A requirement
Características y beneficios
  • Specially formulated phenolic resin and liner adhesive to withstand the effects of repeated autoclaving
  • PTFE faced/general purpose white rubber liners are highly resistant to chemical effects
  • Excellent for general laboratory use
  • Identified in ASTM Specification E982, Class A requirements
Información legal
KIMBLE is a registered trademark of DWK Life Sciences
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)