Documentos clave
DWK45066C-13
KIMBLE® Solid Cap
black phenolic cap (with PTFE Liner), cap for 13-415
Seleccione un Tamaño
300 EA
US$ 594,00
US$ 594,00
PRECIOS SIN IMPUESTOS NACIONALES
Check Cart for Availability
Seleccione un Tamaño
Cambiar Vistas
300 EA
US$ 594,00
About This Item
Código UNSPSC:
41121800
NACRES:
NB.11
US$ 594,00
PRECIOS SIN IMPUESTOS NACIONALES
Check Cart for Availability
Materiales
PTFE/rubber liner
black phenolic cap (with PTFE Liner)
esterilidad
non-sterile
Características
class B
envase
case of 300 ea
fabricante / nombre comercial
Kimble® 45066C-13
Adaptador
cap for 13-415
Descripción general
Class B Caps with PTFE-Faced Rubber Liners highly resistant to temperature and steam during autoclaving
  • Specially formulated phenolic cap material
  • Autoclavable
  • White rubber liners
  • Excellent for general laboratory use
  • Identified in ASTM Specification E982, Class B requirement
Características y beneficios
  • Specially formulated phenolic resin and liner adhesive to withstand the effects of repeated autoclaving
  • PTFE faced/general purpose white rubber liners are highly resistant to chemical effects
  • Excellent for general laboratory use
  • Identified in ASTM Specification E982, Class A requirements
Información legal
KIMBLE is a registered trademark of DWK Life Sciences
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)