Merck
713236
Sigma-Aldrich
(9R)-6′-Methoxycinchonan-9-amine trihydrochloride
≥90.0%
Sinónimos:
(9R)-9-Amino-9-deoxyquinidine trihydrochloride, (R)-(6-Methoxyquinolin-4-yl)((2R,4S,5R)-5-vinylquinuclidin-2-yl)methanamine, Quinine-9-epiamine
About This Item
Fórmula empírica (notación de Hill):
C20H25N3O · 3HCl
Número de CAS:
Peso molecular:
432.81
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22
assay
≥90.0% (HPLC)
≥90.0%
90.0-110.0% (AT)
form
lumps
optical activity
[α]/D +4.0±1.0°, c = 1 in 0.1 M HCl
SMILES string
Cl.Cl.Cl.COc1ccc2nccc([C@@H](N)C3C[C@@H]4CCN3C[C@@H]4C=C)c2c1
InChI
1S/C20H25N3O.3ClH/c1-3-13-12-23-9-7-14(13)10-19(23)20(21)16-6-8-22-18-5-4-15(24-2)11-17(16)18;;;/h3-6,8,11,13-14,19-20H,1,7,9-10,12,21H2,2H3;3*1H/t13-,14-,19?,20+;;;/m0.../s1
InChI key
GZXDEKGNTLNSGM-BQIOMSPYSA-N
Packaging
Bottomless glass bottle. Contents are inside inserted fused cone.
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Visite la Librería de documentos
Los clientes también vieron
Lisa M Sutton et al.
Inflammatory bowel diseases, 20(8), 1324-1328 (2014-07-02)
Lymphocytic esophagitis (LE) is a term recently suggested for the finding of >20 intraepithelial lymphocytes/high-power field in an esophageal biopsy with no more than a rare granulocyte. Two prior studies of LE suggested an association of LE with Crohn's disease
Laith Alkukhun et al.
COPD, 11(6), 670-680 (2014-07-02)
Pulmonary hypertension (PH) is an indicator of poor prognosis in COPD patients; particularly in those with mean pulmonary artery pressure ≥ 40 mm Hg. Electrocardiography (ECG) might be useful for screening of this condition. Retrospective study of COPD patients evaluated
Subashree Mohanraj et al.
International journal of biological macromolecules, 70, 116-123 (2014-07-02)
In the present work, films of pHPMA, pHPMAS hybrid, pHPMA-CPT and pHPMAS-CPT hybrid were prepared by solvent casting method and characterized by FT-IR, FT-Raman, XRD and DSC, respectively. The biocompatibility of the prepared pHPMA film and pHPMAS hybrid film were
Alon Eliakim et al.
Journal of strength and conditioning research, 28(12), 3411-3415 (2014-07-02)
Exercise-associated effects on the growth hormone-insulin-like growth factor-I (GH-IGF-I) axis were studied, mainly after aerobic exercise. We determined the gender effect on the GH-IGF-I axis response to a standard all-out Wingate anaerobic test (WAnT) in healthy active young adult men
Kristy J Kroeker et al.
PloS one, 9(7), e100353-e100353 (2014-07-02)
Ocean acidification (OA) is occurring across a backdrop of concurrent environmental changes that may in turn influence species' responses to OA. Temperature affects many fundamental biological processes and governs key reactions in the seawater carbonate system. It therefore has the