Merck
282812
Sigma-Aldrich
2,2′-Bipyridine-4,4′-dicarboxylic acid
technical grade, 90%
Sinónimos:
2,2′-Bipyridyl-4,4′-dicarboxylic acid
About This Item
Fórmula empírica (notación de Hill):
C12H8N2O4
Número de CAS:
Peso molecular:
244.20
Beilstein/REAXYS Number:
212894
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
grade
technical grade
assay
90%
form
solid
SMILES string
OC(=O)c1ccnc(c1)-c2cc(ccn2)C(O)=O
InChI
1S/C12H8N2O4/c15-11(16)7-1-3-13-9(5-7)10-6-8(12(17)18)2-4-14-10/h1-6H,(H,15,16)(H,17,18)
InChI key
FXPLCAKVOYHAJA-UHFFFAOYSA-N
Other Notes
remainder monomethyl ester
pictograms
Exclamation mark
signalword
Warning
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)
Visite la Librería de documentos
Bo Zheng et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(30), E3987-E3996 (2015-06-28)
New dyads consisting of a strongly absorbing Bodipy (dipyrromethene-BF2) dye and a platinum diimine dithiolate (PtN2S2) charge transfer (CT) chromophore have been synthesized and studied in the context of the light-driven generation of H2 from aqueous protons. In these dyads
Xiang Huang et al.
The Analyst, 140(17), 6015-6024 (2015-07-21)
An efficient aptasensor was developed in which graphene oxide (GO) was employed as an indicator for both electrochemical impedance spectroscopy and electrochemiluminescence (ECL) signal generation. The aptasensor was fabricated by self-assembling the ECL probe of a thiolated adenosine triphosphate binding
Nathan R Rose et al.
Journal of medicinal chemistry, 51(22), 7053-7056 (2008-10-24)
The dynamic methylation of histone lysyl residues plays an important role in biology by regulating transcription, maintaining genomic integrity, and by contributing to epigenetic effects. Here we describe a variety of inhibitor scaffolds that inhibit the human 2-oxoglutarate-dependent JMJD2 subfamily
Maxence Urbani et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 21(45), 16252-16265 (2015-09-12)
The detailed synthesis and characterization of four ruthenium(II) complexes [RuLL'(NCS)2 ] is reported, in which L represents a 2,2'-bipyridine ligand functionalized at the 4,4' positions with benzo[1,2-b:4,5-b']dithiophene derivatives (BDT) and L' is 2,2'-bipyridine-4,4'-dicarboxylic acid unit (dcbpy) (NCS=isothiocyanate). The reaction conditions