Merck
106534
Sigma-Aldrich
1,1,2,2-Tetrachloroethane
reagent grade, 97%
Sinónimos:
Acetylene tetrachloride
About This Item
Fórmula lineal:
CHCl2CHCl2
Número de CAS:
Peso molecular:
167.85
Beilstein/REAXYS Number:
969206
EC Number:
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
grade
reagent grade
vapor density
5.8 (vs air)
vapor pressure
8 mmHg ( 20 °C)
assay
97%
form
liquid
refractive index
n20/D 1.494 (lit.)
bp
147 °C (lit.)
mp
−43 °C (lit.)
density
1.586 g/mL at 25 °C (lit.)
SMILES string
ClC(Cl)C(Cl)Cl
InChI
1S/C2H2Cl4/c3-1(4)2(5)6/h1-2H
InChI key
QPFMBZIOSGYJDE-UHFFFAOYSA-N
pictograms
Skull and crossbonesEnvironment
signalword
Danger
Hazard Classifications
Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Aquatic Chronic 2
Storage Class
6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Visite la Librería de documentos
Wan-Chun Tsai et al.
Journal of chromatography. A, 1216(45), 7846-7850 (2009-10-07)
A novel sample preparation method "Dispersive liquid-liquid-liquid microextraction" (DLLLME) was developed in this study. DLLLME was combined with liquid chromatography system to determine chlorophenoxy acid herbicide in aqueous samples. DLLLME is a rapid and environmentally friendly sample pretreatment method. In
Srigokul Upadhyayula et al.
The journal of physical chemistry. B, 115(30), 9473-9490 (2011-06-21)
Electrostatic properties of proteins are crucial for their functionality. Carboxyamides are small polar groups that, as peptide bonds, are principal structural components of proteins that govern their electrostatic properties. We investigated the medium dependence of the molar polarization and of
Martin Elsner et al.
Environmental science & technology, 41(11), 4111-4117 (2007-07-07)
Despite widespread implementation of zero-valent iron remediation schemes, the manner and order of chemical bond cleavage in iron-mediated organohalide transformations remains imperfectly understood. We present insights from carbon isotope fractionation for the dehalogenation of 1,1,2,2-tetrachloroethane (1,1,2,2-TeCA) and 1,1,1-trichloroethane (1,1,1-TCA) by
William A Arnold et al.
Environmental science & technology, 36(16), 3536-3541 (2002-09-07)
Products of the transformation of organic pollutants in the environment are often predicted based on the structure of the parent compounds. In some cases, however, multiple products may result from the same reaction pathway. In this study, the reduction of
Debolina Basu et al.
Bioresource technology, 101(1), 21-25 (2009-08-25)
This study examines the performance of bench-scale Upflow Anaerobic Sludge Blanket (UASB) reactor in treating simulated wastewaters containing 1,1,2,2-tetrachloroethane (TeCA). Reactors R1 (control) and R2 (containing TeCA) were operated at Hydraulic Retention Times (HRTs) of 36, 30, 24, 18 and