Merck
  • ERK5 signalling pathway is essential for fluid shear stress-induced COX-2 gene expression in MC3T3-E1 osteoblast.
ERK5 signalling pathway is essential for fluid shear stress-induced COX-2 gene expression in MC3T3-E1 osteoblast.
Molecular and cellular biochemistry (2015-05-16)
Jin Jiang, Liang-gong Zhao, Yuan-jun Teng, Shao-long Chen, Li-ping An, Jing-ling Ma, Jing Wang, Ya-yi Xia
ABSTRACT
Bone cells respond to various mechanical stimuli including fluid shear stress (FSS) in vitro. Induction of cyclooxygenase-2 (COX-2) is thought to be important for the anabolic effects of mechanical loading. Recently, extracellular-signal-regulated kinase 5 (ERK5) has been found to be involved in multiple cellular processes. However, the relationship between ERK5 and the induction of COX-2 is still unknown. Here, we investigated the potential involvement of ERK5 in the response of pre-osteoblastic MC3T3-E1 cells upon FSS. MC3T3-E1 cells were subjected to 12 dyn/cm(2) FSS. Then, we established a ERK5 small interfering RNA (siRNA) transfected cell line using the MC3T3-E1 cells. After the successful transfection confirmed by real-time reverse transcription-polymerase chain reaction and Western blotting, the expression of COX-2, cAMP response element-binding protein (CREB), and nuclear factor kappa B cells (NF-κB) were assayed for downstream effectors of activated ERK5 under FSS by Western blotting. Our results showed that FSS could stimulate COX-2 activity, and induce the phosphorylation of ERK5, CREB, and NF-κB. When the MC3T3-E1 cells were transfected using siRNA before exposure to FSS, COX-2 activity was suppressed, and the phosphorylation of CREB and NF-κB was significantly downregulated. In summary, we demonstrated that ERK5 pathway is essential in the induction of COX-2 gene.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Mapk7
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
MISSION® esiRNA, targeting human MAPK7
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)