Merck
  • Concave microwell array-mediated three-dimensional tumor model for screening anticancer drug-loaded nanoparticles.
Concave microwell array-mediated three-dimensional tumor model for screening anticancer drug-loaded nanoparticles.
Nanomedicine : nanotechnology, biology, and medicine (2015-03-11)
AhRan Kang, Hye In Seo, Bong Geun Chung, Sang-Hoon Lee
ABSTRACT
We investigated the effect of anticancer drug-loaded functional polymeric nanoparticles on drug resistance of three-dimensional (3D) breast tumor spheroids. 3D tumor models were built using concave microwells with different diameters (300-700μm) and nanoparticles were prepared using thermo-responsive poly(N-isopropylacrylamide) (PNIPAM)-co-acrylic acid (AA). Upon culturing with doxorubicin-loaded PNIPAM-co-AA nanoparticles for 96hours, the smallest tumor spheroids were extensively disrupted, resulting in a reduction in spheroid diameter. In contrast, the sizes of the largest tumor spheroids were not changed. Scanning electron microscopy revealed that the circular shape of 3D spheroids treated with doxorubicin-loaded PNIPAM-co-AA nanoparticles had collapsed severely. Cell viability assays also demonstrated that the largest tumor spheroids cultured with doxorubicin-loaded PNIPAM-co-AA nanoparticles were highly resistant to the anticancer drug. We confirmed that tight cell-cell contacts within largest tumor spheroids significantly improved the anticancer drug resistance. Therefore, this uniform-sized 3D breast tumor model could be a potentially powerful tool for anticancer drug screening applications. The battle against cancer is a big challenge. With new anti-cancer drugs being developed under the nanotechnology platform, there is a need to have a consistent and reliable testing system that mimics the in-vivo tumor scenario. The authors successfully designed a 3D tumor model using concave microwells to produce different tumor diameters. This will be of value for future drug screening.
Sigma-Aldrich
N-Isopropylacrylamide, 97%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 mL
Please contact Customer Service for Availability
CA$46.70
50 mL
Please contact Customer Service for Availability
CA$95.10
100 mL
Please contact Customer Service for Availability
CA$176.00
Sigma-Aldrich
N-Isopropylacrylamide, ≥99%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 mL
Please contact Customer Service for Availability
CA$46.70
50 mL
Please contact Customer Service for Availability
CA$95.10
100 mL
Please contact Customer Service for Availability
CA$176.00
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 mL
Please contact Customer Service for Availability
CA$46.70
50 mL
Please contact Customer Service for Availability
CA$95.10
100 mL
Please contact Customer Service for Availability
CA$176.00
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 mL
Please contact Customer Service for Availability
CA$46.70
50 mL
Please contact Customer Service for Availability
CA$95.10
100 mL
Please contact Customer Service for Availability
CA$176.00
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 mL
Please contact Customer Service for Availability
CA$46.70
50 mL
Please contact Customer Service for Availability
CA$95.10
100 mL
Please contact Customer Service for Availability
CA$176.00
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 mL
Please contact Customer Service for Availability
CA$46.70
50 mL
Please contact Customer Service for Availability
CA$95.10
100 mL
Please contact Customer Service for Availability
CA$176.00
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 mL
Please contact Customer Service for Availability
CA$46.70
50 mL
Please contact Customer Service for Availability
CA$95.10
100 mL
Please contact Customer Service for Availability
CA$176.00
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 mL
Please contact Customer Service for Availability
CA$46.70
50 mL
Please contact Customer Service for Availability
CA$95.10
100 mL
Please contact Customer Service for Availability
CA$176.00
Sigma-Aldrich
Calcein-AM, Small Package (20 X 50 μg ), ≥95.0% (HPLC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 mL
Please contact Customer Service for Availability
CA$46.70
50 mL
Please contact Customer Service for Availability
CA$95.10
100 mL
Please contact Customer Service for Availability
CA$176.00
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 mL
Please contact Customer Service for Availability
CA$46.70
50 mL
Please contact Customer Service for Availability
CA$95.10
100 mL
Please contact Customer Service for Availability
CA$176.00
Sigma-Aldrich
Calcein-AM, suitable for fluorescence, BioReagent, ≥90% (HPLC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 mL
Please contact Customer Service for Availability
CA$46.70
50 mL
Please contact Customer Service for Availability
CA$95.10
100 mL
Please contact Customer Service for Availability
CA$176.00
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 mL
Please contact Customer Service for Availability
CA$46.70
50 mL
Please contact Customer Service for Availability
CA$95.10
100 mL
Please contact Customer Service for Availability
CA$176.00
Sigma-Aldrich
Sucrose, ACS reagent
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 mL
Please contact Customer Service for Availability
CA$46.70
50 mL
Please contact Customer Service for Availability
CA$95.10
100 mL
Please contact Customer Service for Availability
CA$176.00
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 mL
Please contact Customer Service for Availability
CA$46.70
50 mL
Please contact Customer Service for Availability
CA$95.10
100 mL
Please contact Customer Service for Availability
CA$176.00
Sigma-Aldrich
Sucrose, for molecular biology, ≥99.5% (GC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 mL
Please contact Customer Service for Availability
CA$46.70
50 mL
Please contact Customer Service for Availability
CA$95.10
100 mL
Please contact Customer Service for Availability
CA$176.00
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 mL
Please contact Customer Service for Availability
CA$46.70
50 mL
Please contact Customer Service for Availability
CA$95.10
100 mL
Please contact Customer Service for Availability
CA$176.00
Sigma-Aldrich
Sucrose, Grade I, ≥99% (GC), suitable for plant cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 mL
Please contact Customer Service for Availability
CA$46.70
50 mL
Please contact Customer Service for Availability
CA$95.10
100 mL
Please contact Customer Service for Availability
CA$176.00
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 mL
Please contact Customer Service for Availability
CA$46.70
50 mL
Please contact Customer Service for Availability
CA$95.10
100 mL
Please contact Customer Service for Availability
CA$176.00
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 mL
Please contact Customer Service for Availability
CA$46.70
50 mL
Please contact Customer Service for Availability
CA$95.10
100 mL
Please contact Customer Service for Availability
CA$176.00
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 mL
Please contact Customer Service for Availability
CA$46.70
50 mL
Please contact Customer Service for Availability
CA$95.10
100 mL
Please contact Customer Service for Availability
CA$176.00
Sigma-Aldrich
Sucrose, BioUltra, for molecular biology, ≥99.5% (HPLC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 mL
Please contact Customer Service for Availability
CA$46.70
50 mL
Please contact Customer Service for Availability
CA$95.10
100 mL
Please contact Customer Service for Availability
CA$176.00
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 mL
Please contact Customer Service for Availability
CA$46.70
50 mL
Please contact Customer Service for Availability
CA$95.10
100 mL
Please contact Customer Service for Availability
CA$176.00
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 mL
Please contact Customer Service for Availability
CA$46.70
50 mL
Please contact Customer Service for Availability
CA$95.10
100 mL
Please contact Customer Service for Availability
CA$176.00
Sigma-Aldrich
Calcein AM solution, 4 mM in DMSO, ≥90% (HPLC), solution
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
20 mL
Please contact Customer Service for Availability
CA$46.70
50 mL
Please contact Customer Service for Availability
CA$95.10
100 mL
Please contact Customer Service for Availability
CA$176.00