Skip to Content
Merck
All Photos(2)

Key Documents

S5391

Sigma-Aldrich

Sucrose

≥99.5% (GC), Grade II, suitable for plant cell culture

Synonym(s):

α-D-Glc-(1→2)-β-D-Fru, α-D-Glucopyranosyl β-D-fructofuranoside, β-D-Fructofuranosyl-α-D-glucopyranoside, D(+)-Saccharose, Sugar

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
$49.80
5 KG
$175.00
25 KG
$1,460.00

$49.80

PRICE WITHOUT NATIONAL TAXES

Available to ship onApril 03, 2025Details


Request a Bulk Order

Select a Size

Change View
1 KG
$49.80
5 KG
$175.00
25 KG
$1,460.00

About This Item

Empirical Formula (Hill Notation):
C12H22O11
CAS Number:
Molecular Weight:
342.30
Beilstein:
90825
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.28

$49.80

PRICE WITHOUT NATIONAL TAXES

Available to ship onApril 03, 2025Details


Request a Bulk Order

biological source

sugar cane

type

Grade II

Assay

≥99.5% (GC)

form

powder

availability

not available in Japan

technique(s)

cell culture | plant: suitable

color

white

useful pH range

5.5-7 (25 °C, 342 g/L)

mp

185-187 °C (lit.)

solubility

H2O: 0.5 g/mL, clear, colorless to faintly yellow

application(s)

agriculture

storage temp.

room temp

SMILES string

OC[C@H]1O[C@H](O[C@]2(CO)O[C@H](CO)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C12H22O11/c13-1-4-6(16)8(18)9(19)11(21-4)23-12(3-15)10(20)7(17)5(2-14)22-12/h4-11,13-20H,1-3H2/t4-,5-,6-,7-,8+,9-,10+,11-,12+/m1/s1

InChI key

CZMRCDWAGMRECN-UGDNZRGBSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Use in plant cell culture applications as a cryopreservative.

Other Notes

Slightly less pure than Grade I
To gain a comprehensive understanding of our extensive range of Disaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Denise Harrison et al.
Pediatrics, 130(5), 918-925 (2012-10-10)
The use of oral sucrose has been the most extensively studied pain intervention in newborn care to date. More than 150 published studies relating to sweet-taste-induced calming and analgesia in human infants have been identified, of which 100 (65%) include
Rafaela Ribeiro Reis et al.
Plant science : an international journal of experimental plant biology, 221-222, 59-68 (2014-03-25)
Drought is one of the most challenging agricultural issues limiting sustainable sugarcane production and, in some cases, yield losses caused by drought are nearly 50%. DREB proteins play vital regulatory roles in abiotic stress responses in plants. The transcription factor
Zsuzsanna Tucsek et al.
The journals of gerontology. Series A, Biological sciences and medical sciences, 69(10), 1212-1226 (2013-11-26)
There is growing evidence that obesity has deleterious effects on the brain and cognitive function in the elderly population. However, the specific mechanisms through which aging and obesity interact to promote cognitive decline remain unclear. To test the hypothesis that
Shlomi Cohen et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 40(3), 774-790 (2014-09-23)
The hypothalamic-pituitary-adrenal (HPA) axis displays a characteristic circadian pattern of corticosterone release, with higher levels at the onset of the active phase and lower levels at the onset of the inactive phase. As corticosterone levels modify the response to stress
William J Armour et al.
The Plant cell, 27(9), 2484-2500 (2015-08-25)
Lobe development in the epidermal pavement cells of Arabidopsis thaliana cotyledons and leaves is thought to take place via tip-like growth on the concave side of lobes driven by localized concentrations of actin filaments and associated proteins, with a predicted

Questions

  1. How long does sucrose water stay stable before it goes bad or becomes moldy?

    1 answer
    1. There is no available information or data on the stability duration of reconstituted sucrose water for mice consumption. This specific data has not been tested.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service