Merck
Permeation of Dopamine Sulfate through the Blood-Brain Barrier.
PloS one (2015-07-25)
Tina Suominen, T Petteri Piepponen, Risto Kostiainen
ABSTRACT
Dopamine sulfate (DA-3- and DA-4-S) have been determined in the human brain, but it is unclear whether they are locally formed in the central nervous system (CNS), or transported into the CNS from peripheral sources. In the current study, permeation of the blood-brain barrier (BBB) by DA-S was studied by injecting 13C6-labelled regioisomers of DA-S (13DA-3-S and 13DA-4-S) and dopamine (DA) subcutaneously (s.c.) in anesthetized rats, then analyzing brain microdialysis and plasma samples by UPLC-MS/MS. The results in the microdialysis samples demonstrated that brain concentrations of 13DA-S regioisomers clearly increased after the s.c. injections. The concentration of DA did not change, indicating the permeation of DA-S through an intact BBB. The analysis of plasma samples, however, showed that DA-S only permeates the BBB to a small extent, as the concentrations in plasma were substantially higher than in the microdialysis samples. The results also showed that the concentrations of DA-3-S were around three times higher than the concentrations of DA-4-S in rat brain, as well as in the plasma samples after the s.c. injections, indicating that DA-3-S and DA-4-S permeate the BBB with similar efficiency. The fate of 13DA-S in brain was followed by monitoring 13C6-labelled DA-S hydrolysis products, i.e. 13DA and its common metabolites; however, no 13C6-labelled products were detected. This suggests that DA-S either permeates through the BBB back to the peripheral circulation or is dissociated or metabolized by unexpected mechanisms.
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
Potassium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, anhydrous, 99.9 atom % D
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
3,4-Dihydroxyphenylacetic acid, 98%
Sigma-Aldrich
Dimethyl sulfoxide-d6, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Potassium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
Potassium chloride, ≥99.99% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.9 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Potassium chloride, 99.999% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide-d6, 99.5 atom % D
Sigma-Aldrich
Dimethyl sulfoxide-d6, "Special HOH", ≥99.9 atom % D
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
Dopamine hydrochloride
Sigma-Aldrich
Potassium chloride, for molecular biology, ≥99.0%
Sigma-Aldrich
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Potassium chloride solution, 0.075 M, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Potassium chloride, BioXtra, ≥99.0%
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%