Skip to Content
Merck
All Photos(3)

Key Documents

409316

Sigma-Aldrich

Potassium chloride

99.999% trace metals basis

Synonym(s):

Potassium monochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KCl
CAS Number:
Molecular Weight:
74.55
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.999% trace metals basis

form

solid

technique(s)

protein extraction: suitable

impurities

≤15.0 ppm Trace Metal Analysis

mp

770 °C (lit.)

density

1.98 g/mL at 25 °C (lit.)

SMILES string

[Cl-].[K+]

InChI

1S/ClH.K/h1H;/q;+1/p-1

InChI key

WCUXLLCKKVVCTQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Potassium chloride (KCl) is a water-soluble metal salt that comprises of potassium and chlorine. It can be extracted from minerals and salt water. KCl can be used in industries such as cosmetics, food, biomedical, chemical and fertilizer.

Application

May be used for the preparation of phosphate buffered saline, and for the extraction and solubilization of proteins.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Potassium Chloride for Improved Urea Fertilizer Efficiency 1
Rappaport BD and Axley JH
Soil Science Society of America Journal. Soil Science Society of America, 48(2), 399-401 (1984)
Potassium compounds
Freilich MB, et al.
Kirk-Othmer Encyclopedia of Chemical Technology, 1-35 (2000)
Synthesis and characterization of bioactive hydroxyapatite-calcite nanocomposite for biomedical applications
Kumar GS, et al.
Journal of Colloid and Interface Science, 349(1), 56-62 (2010)
Use of gelatin in the differential-pulse polarographic determination and identification of synthetic colouring matters in drugs and cosmetics
Barros AA, et al.
Analyst, 113(6), 853-858 (1988)
Calista K L Ng et al.
Human molecular genetics, 24(11), 3163-3171 (2015-02-26)
mRNA decay is an essential and active process that allows cells to continuously adapt gene expression to internal and environmental cues. There are two mRNA degradation pathways: 3' to 5' and 5' to 3'. The DCPS protein is the scavenger

Articles

Magnetic nanoparticles have attracted tremendous attention due to their novel properties and their potential applications in magnetic recording, magnetic energy storage and biomedicine.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service