Skip to Content
Merck
All Photos(1)

Key Documents

850217

Sigma-Aldrich

3,4-Dihydroxyphenylacetic acid

98%

Synonym(s):

DOPAC, Homoprotocatechuic acid

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$59.60
5 G
$274.00

$59.60

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$59.60
5 G
$274.00

About This Item

Linear Formula:
(HO)2C6H3CH2CO2H
CAS Number:
Molecular Weight:
168.15
Beilstein:
2211017
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$59.60

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

powder

mp

127-130 °C (lit.)

solubility

water: soluble 50 mg/mL, clear, almost colorless

functional group

carboxylic acid

SMILES string

OC(=O)Cc1ccc(O)c(O)c1

InChI

1S/C8H8O4/c9-6-2-1-5(3-7(6)10)4-8(11)12/h1-3,9-10H,4H2,(H,11,12)

InChI key

CFFZDZCDUFSOFZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3,4-dihydroxyphenylacetic acid (DOPAC) is a normal constituent of rat brain tissue.[1] A mass fragmentographic method for determination of DOPAC in rat brain tissue has been described.[1] It is major metabolite of dopamine (DA).[2] The voltammetric reduction of DOPAC has been studied at a glassy carbon electrode modified with single-wall carbon nanotubes (SWNTs).[3] Semi-automatic fluorometric assay technique for DOPAC has been reported.[4]

Application

3,4-Dihydroxyphenylacetic acid may be employed as ligand in the preparation of Mo(VI) complexes.[5]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Y Deutch et al.
Brain research, 333(1), 143-146 (1985-04-29)
The effects of stress on dopamine (DA) metabolism in the mesencephalic DA cell body areas and DA terminal field regions were examined. Both mild footshock stress and exposure to a neutral stimulus previously paired with footshock resulted in a selective
Simultaneous mass fragmentographic determination of 3, 4-dihydroxyphenylacetic acid and 4 hydroxy-3-methoxyphenylacetic acid in brain tissue.
F A Wiesel et al.
Journal of neural transmission, 35(4), 319-326 (1974-01-01)
Molybdenum catecholates as models for Mo in biological systems. 1. Synthesis and spectroscopic study on Mo complexes with 3, 4-dihydroxybenzoic and 3, 4-dihydroxyphenylacetic acid.
Karaliota A, et al.
Journal of Inorganic Biochemistry, 69(1), 79-90 (1998)
B H Westerink et al.
European journal of pharmacology, 38(2), 281-291 (1976-08-01)
A concurrent semi-automatic fluorometric assay technique for 3,4-dihydroxyphenylacetic acid (DOPAC) and homovanillic acid (HVA), is described. The method is based on a rapid manually performed isolation of DOPAC and HVA on small columns of Sephadex G-10 followed by an automated
Electrocatalytic oxidation of 3, 4-dihydroxyphenylacetic acid at a glassy carbon electrode modified with single-wall carbon nanotubes.
Wang J, et al.
Electrochimica Acta, 47(4), 651-657 (2001)

Questions

1–3 of 3 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. Can it be stored at -20C

    1 answer
    1. This product is stored at room temperature. In general, powder materials are not affected by freezer storage, however, this item has not been tested under these storage conditions.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service