Merck
  • In-line coupling of single-drop microextraction with capillary electrophoresis-mass spectrometry.
In-line coupling of single-drop microextraction with capillary electrophoresis-mass spectrometry.
Analytical and bioanalytical chemistry (2015-09-26)
Jihye Kim, Kihwan Choi, Doo Soo Chung
ABSTRACT
Single-drop microextraction (SDME) was in-line coupled with capillary electrophoresis-mass spectrometry to provide sample cleanup and enrichment simultaneously. Since there is no outlet vial in a conventional capillary electrophoresis-electrospray ionization-mass spectrometry (CE-ESI-MS) configuration, it is not easy to hang a single drop in the capillary inlet for extraction. We overcame the difficulty of coupling SDME and CE-MS by using a temporary outlet reservoir. Basic drugs such as methamphetamine, amphetamine, phenethylamine, methoxyphenamine, and mephentermine were extracted from a basic sample solution to an acidic acceptor drop covered with a thin octanol layer formed at the capillary inlet tip. Compared to the CE-MS method in the multiple reaction monitoring (MRM) mode, the in-line SDME-CE-MS/MS technique showed 130∼150-fold enrichment in 10 min. The relative standard deviations (RSDs) of peak height ranged from 9 to 13 %. RSDs can be reduced from 4 to 6 % using mephentermine as an internal standard. We examined the pretreatment of sample with and without SDME from human urine under the full-scan mode, which confirmed that many metabolites were cleaned up by the selective extraction method of SDME. Even if the analytes from human urine were analyzed under the MRM mode used as a mass filter, there was an isobaric compound causing a disturbance to the analysis. However, in-line SDME-CE-MS/MS made it possible to perform a sample cleanup as well as sample enrichment. The research is extremely advantageous in that it is rapid, convenient, and highly sensitive for the analysis of biological samples using a commercially available instrument.
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
Trimethoxy(octadecyl)silane, technical grade
Sigma-Aldrich
Phenethylamine, purified by redistillation, ≥99.5%
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Phenethylamine, ≥99%
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Sigma-Aldrich
N-Benzylmethylamine, 97%
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Hippuric acid, 98%
Sigma-Aldrich
Phenethylamine, 99%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Sodium hydroxide, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%