Merck
  • Validation of 64Cu-DOTA-rituximab injection preparation under good manufacturing practices: a PET tracer for imaging of B-cell non-Hodgkin lymphoma.
Validation of 64Cu-DOTA-rituximab injection preparation under good manufacturing practices: a PET tracer for imaging of B-cell non-Hodgkin lymphoma.
Molecular imaging (2015-03-13)
Arutselvan Natarajan, Natasha Arksey, Andrei Iagaru, Frederick T Chin, Sanjiv Sam Gambhir
ABSTRACT
Manufacturing of 64Cu-1,4,7,10-tetraazacyclododecane-N,N',N'',N'''-tetraacetic acid (DOTA)-rituximab injection under good manufacturing practices (GMP) was validated for imaging of patients with CD20+ B-cell non-Hodgkin lymphoma. Rituximab was purified by size exclusion high performance liquid chromatography (HPLC) and conjugated to DOTA-mono-(N-hydroxysuccinimidyl) ester. 64CuCl2, buffers, reagents, and other raw materials were obtained as high-grade quality. Following a semi-automated synthesis of 64Cu-DOTA-rituximab, a series of quality control tests was performed. The product was further tested in vivo using micro-positron emission tomography/computed tomography (PET/CT) to assess targeting ability towards human CD20 in transgenic mice. Three batches of 64Cu-DOTA-rituximab final product were prepared as per GMP specifications. The radiolabeling yield from these batches was 93.1 ± 5.8%; these provided final product with radiopharmaceutical yield, purity, and specific activity of 59.2 ± 5.1% (0.9 ± 0.1 GBq of 64Cu), > 95% (by HPLC and radio-thin layer chromatography), and 229.4 ± 43.3 GBq/µmol (or 1.5 ± 0.3 MBq/µg), respectively. The doses passed apyrogenicity and human serum stability specifications, were sterile up to 14 days, and retained > 60% immunoreactivity. In vivo micro-PET/CT mouse images at 24 hours postinjection showed that the tracer targeted the intended sites of human CD20 expression. Thus, we have validated the manufacturing of GMP grade 64Cu-DOTA-rituximab for injection in the clinical setting.
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FG
Sigma-Aldrich
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Sinapic acid, ≥98%, powder
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Trisodium citrate dihydrate, meets USP testing specifications
Sigma-Aldrich
Diethylenetriaminepentaacetic acid, ≥98% (titration)
Sigma-Aldrich
Sodium acetate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium acetate, anhydrous, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium acetate, puriss. p.a., ACS reagent, reag. Ph. Eur., anhydrous
Sigma-Aldrich
Sodium acetate, ACS reagent, ≥99.0%
Sigma-Aldrich
Diethylenetriaminepentaacetic acid, ≥99% (titration)
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, puriss., 99-100%
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Acetic acid, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.8-100.5%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Tantalum(V) ethoxide, 99.98% trace metals basis
Sigma-Aldrich
Sodium citrate tribasic dihydrate, puriss. p.a., ACS reagent, ≥99.0% (NT)