Skip to Content
Merck
All Photos(4)

Key Documents

S7545

Sigma-Aldrich

Sodium acetate

BioXtra, ≥99.0%

Synonym(s):

Acetic acid sodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
$68.90
500 G
$124.00
1 KG
$241.00

$68.90


Estimated to ship onMay 25, 2025


Request a Bulk Order

Select a Size

Change View
250 G
$68.90
500 G
$124.00
1 KG
$241.00

About This Item

Linear Formula:
CH3COONa
CAS Number:
Molecular Weight:
82.03
Beilstein:
3595639
EC Number:
MDL number:
UNSPSC Code:
12161700
eCl@ss:
39021906
PubChem Substance ID:
NACRES:
NA.25

$68.90


Estimated to ship onMay 25, 2025


Request a Bulk Order

product line

BioXtra

Quality Level

Assay

≥99.0%

form

solid

autoignition temp.

1112 °F

impurities

≤0.0005% Phosphorus (P)
<0.01% Insoluble matter

pH

8.5-9.9 (25 °C, 246 g/L)

pKa 

4.76 (acetic acid)

mp

>300 °C (dec.) (lit.)

solubility

H2O: 1 M, clear, colorless

anion traces

chloride (Cl-): ≤0.002%
sulfate (SO42-): ≤0.003%

cation traces

Al: ≤0.0005%
Ca: ≤0.005%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.03%
Mg: ≤0.0005%
NH4+: ≤0.05%
Pb: ≤0.001%
Zn: ≤0.0005%

SMILES string

[Na+].CC([O-])=O

InChI

1S/C2H4O2.Na/c1-2(3)4;/h1H3,(H,3,4);/q;+1/p-1

InChI key

VMHLLURERBWHNL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium acetate is commonly applicable in molecular biology experiments such as nucleic acid purification and precipitation, protein gel electrophoresis, protein crystallization, and high-performance liquid chromatography (HPLC). Sodium acetate is also used in plastic production, dyeing, and leather tanning.[1]

Application

Sodium acetate has been used:
  • in the isolation of DNA from bacterial samples of the human gastrointestinal tract[2]
  • as a component of RNA elution buffer for the preparation of infrared (IR) fluorescent probes in northern blot technique[3]
  • in the lysis buffer for cellular lysis to detect reversible oxidation of protein tyrosine phosphatases (PTPs) with the cysteinyl-labeling assay using IMR90 fibroblasts[4]

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Density, viscosity and ultrasonic velocity studies of aqueous solutions of sodium acetate at different temperatures
Kharat SJ
Journal of Molecular Liquids, 140(1-3), 10-14 (2008)
Christopher Fields et al.
Bio-protocol, 9(8) (2019-08-21)
Northern blot is a molecular biology technique that can detect, quantify, and determine the molecular weight of RNA. Recently, we published a protocol utilizing near-infrared (IR) fluorescent probes in Northern blot (irNorthern). Our method is as sensitive as other non-radioactive
Erwin G Zoetendal et al.
Nature protocols, 1(2), 870-873 (2007-04-05)
The human gastrointestinal (GI) tract contains a complex microbial community that develops in time and space. The most widely used approaches to study microbial diversity and activity are all based on the analysis of nucleic acids, DNA, rRNA and mRNA.
Amanda P De Souza et al.
Journal of experimental botany, 66(14), 4351-4365 (2015-04-25)
The production of bioenergy from grasses has been developing quickly during the last decade, with Miscanthus being among the most important choices for production of bioethanol. However, one of the key barriers to producing bioethanol is the lack of information
Graham R Scott et al.
Molecular biology and evolution, 32(8), 1962-1976 (2015-04-09)
At high-altitude, small mammals are faced with the energetic challenge of sustaining thermogenesis and aerobic exercise in spite of the reduced O2 availability. Under conditions of hypoxic cold stress, metabolic demands of shivering thermogenesis and locomotion may require enhancements in

Questions

  1. 시약이 solid라고 되어 있는데 solid가 무슨 이미인가요? 시약이 알맹이 형태로 되어 있는 건가요?

    1 answer
    1. The form of this product is a white powder.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service