Skip to Content
Merck
All Photos(7)

Key Documents

791741

Sigma-Aldrich

Sodium acetate

anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%

Synonym(s):

Acetic acid sodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$87.80
500 G
$134.00
1 KG
$238.00
2.5 KG
$431.00

$87.80


Estimated to ship onMarch 27, 2025


Request a Bulk Order

Select a Size

Change View
100 G
$87.80
500 G
$134.00
1 KG
$238.00
2.5 KG
$431.00

About This Item

Linear Formula:
CH3COONa
CAS Number:
Molecular Weight:
82.03
Beilstein:
3595639
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.21

$87.80


Estimated to ship onMarch 27, 2025


Request a Bulk Order

grade

ACS reagent
anhydrous

Quality Level

product line

Redi-Dri

Assay

≥99.0%

form

solid

autoignition temp.

1112 °F

quality

free-flowing

impurities

≤0.01% insolubles

loss

≤1.0% loss on drying

pH

7.0-9.2 (25 °C, 5%)

pKa 

4.76 (acetic acid)

mp

>300 °C (dec.) (lit.)

anion traces

chloride (Cl-): ≤0.002%
phosphate (PO43-): ≤0.001%
sulfate (SO42-): ≤0.003%

cation traces

Ca: ≤0.005%
Fe: ≤0.001%
Mg: ≤0.002%
heavy metals: ≤0.001% (by ICP)

SMILES string

[Na+].CC([O-])=O

InChI

1S/C2H4O2.Na/c1-2(3)4;/h1H3,(H,3,4);/q;+1/p-1

InChI key

VMHLLURERBWHNL-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium acetate is an inexpensive and non-toxic chemical substance that is widely used in industry as a food additive or as a base for the neutralization of reaction mixtures.

Application

Sodium acetate can be used as a reagent:
  • In the Erlenmeyer-Plochl reaction to synthesize oxazolones by the condensation of aromatic aldehydes and hippuric acid.
  • In palladium-catalyzed arylation of thiophenes using aryl halides.
It can also be used as a base to synthesize cinnamic acids via Perkin reaction.

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sam Joshva Baskar Jesudasan et al.
PloS one, 9(6), e99443-e99443 (2014-06-11)
Microglia are the primary immune cells of the central nervous system (CNS). Membrane bound sensors on their processes monitor the extracellular environment and respond to perturbations of the CNS such as injury or infection. Once activated, microglia play a crucial
Sabina Lukovac et al.
mBio, 5(4) (2014-08-15)
The gut microbiota is essential for numerous aspects of human health. However, the underlying mechanisms of many host-microbiota interactions remain unclear. The aim of this study was to characterize effects of the microbiota on host epithelium using a novel ex
Carol L Ladner-Keay et al.
PloS one, 9(6), e98753-e98753 (2014-06-04)
The formation of β-sheet rich prion oligomers and fibrils from native prion protein (PrP) is thought to be a key step in the development of prion diseases. Many methods are available to convert recombinant prion protein into β-sheet rich fibrils
Mari Endo et al.
Journal of gastroenterology, 49(6), 1026-1039 (2013-07-13)
Daikenchuto (DKT), a gastrointestinal prokinetic Japanese herbal medicine, is prescribed for patients with postoperative ileus (POI) and adhesive bowel obstruction following abdominal surgery. Several mechanisms for the amelioration of POI by DKT have been suggested; however, it has remained unclear
Kyle R Mulligan et al.
European spine journal : official publication of the European Spine Society, the European Spinal Deformity Society, and the European Section of the Cervical Spine Research Society, 24(11), 2395-2401 (2014-09-23)
The aim of the study was to investigate if axial T1ρ MR images had similar accuracy as established sagittal T1ρ MRI for the assessment of proteoglycan concentration and content in intervertebral degenerated discs (IDDs). T1ρ and T2-weighted MR images of

Articles

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service