Merck
  • Towards the chiral metabolomics: Liquid chromatography-mass spectrometry based DL-amino acid analysis after labeling with a new chiral reagent, (S)-2,5-dioxopyrrolidin-1-yl-1-(4,6-dimethoxy-1,3,5-triazin-2-yl)pyrrolidine-2-carboxylate, and the application to saliva of healthy volunteers.
Towards the chiral metabolomics: Liquid chromatography-mass spectrometry based DL-amino acid analysis after labeling with a new chiral reagent, (S)-2,5-dioxopyrrolidin-1-yl-1-(4,6-dimethoxy-1,3,5-triazin-2-yl)pyrrolidine-2-carboxylate, and the application to saliva of healthy volunteers.
Analytica chimica acta (2015-05-06)
Toshiki Mochizuki, Takahiro Takayama, Kenichiro Todoroki, Koichi Inoue, Jun Zhe Min, Toshimasa Toyo'oka
ABSTRACT
A novel triazine-type chiral derivatization reagent, i.e., (S)-2,5-dioxopyrrolidin-1-yl-1-(4,6-dimethoxy-1,3,5-triazin-2-yl) pyrrolidine-2-carboxylate (DMT-(S)-Pro-OSu), was developed for the highly sensitive and selective detection of chiral amines and amino acids by UPLC-MS/MS analysis. The enantiomers of amino acids were easily labeled with the reagents at room temperature within 40 min in an alkaline medium containing triethylamine. The diastereomers derived from proteolytic amino acids, except serine, were well separated under isocratic elution conditions by reversed-phase chromatography using an ODS column (Rs=1.2-9.0). DL-Serine was separated by use of an ADME column which has relatively higher polar surface than the conventional ODS column. The characteristic product ions, i.e., m/z 195.3 and m/z 209.3, were detected from all the diastereomers by the collision-induced dissociation of the protonated molecule. A highly sensitive detection on the amol-fmol level was obtained from the selected reaction monitoring (SRM) chromatogram. The chiral amines (e.g., adrenaline and noradrenaline) labeled with DMT-(S)-Pro-OSu were also well separated and sensitively detected by the present procedure. The method using DMT-(S)-Pro-OSu was used for the determination of DL-amino acids in the human saliva from healthy volunteers. Various L-amino acids were identified in the saliva. Furthermore, D-alanine (D-Ala) and D-proline (D-Pro) were also detected in relatively high concentrations (>5%). The ratio was higher in male saliva than in female saliva. However, the difference in the ratio of D-Ala for one day was not very high and the effect of foods and beverage seemed to be negligible. Based on the results using L-Ala-d3, the D-Ala in saliva seemed to be produced due to the racemization with some enzymes such as racemase. The racemization reaction was reversible, i.e., D-Ala-d3 was also racemized to L-Ala-d3 in saliva. Thus, care should be taken during the analysis of DL-amino acids in saliva. The present method using DMT-(S)-Pro-OSu may be applicable for the determination of chiral amine metabolomics, because the resulting derivatives produce the same product ions without relation to the compounds and show highly sensitive detection in the SRM mode of MS/MS. Consequently, DMT-(S)-Pro-OSu seems to be a useful chiral derivatization reagent for the determination of amines and amino acids in biological samples.
Sigma-Aldrich
Ammonium acetate solution, 7.5 M
Sigma-Aldrich
Ammonium acetate, ≥98%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
(−)-Norepinephrine, ≥98%, crystalline
Sigma-Aldrich
Triethylamine, ≥99%
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Triethylamine, ampule, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, for amino acid analysis, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Triethylamine, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
(R)-(+)-α-Methylbenzylamine, 98%
Sigma-Aldrich
L-Alanine-3,3,3-d3, 99 atom % D
Sigma-Aldrich
Triethylamine, ≥99.5%
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Supelco
Tetrahydrofuran, HPLC grade, ≥99.9%, inhibitor-free
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Methanol
Sigma-Aldrich
(R)-(+)-α-Methylbenzylamine, ChiPros®, produced by BASF, ≥99.0%
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
1,2-Dichloroethane, anhydrous, 99.8%
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Ethylamine solution, 66.0-72.0% in H2O
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Ethylamine solution, 2.0 M in THF