Merck
  • Antibacterial, antioxidant and hypoglycaemic effects of Thymus capitatus (L.) Hoffmanns. et Link leaves' fractions.
Antibacterial, antioxidant and hypoglycaemic effects of Thymus capitatus (L.) Hoffmanns. et Link leaves' fractions.
Journal of enzyme inhibition and medicinal chemistry (2014-07-18)
Liliana Iauk, Rosaria Acquaviva, Silvana Mastrojeni, Andrea Amodeo, Michela Pugliese, Monica Ragusa, Monica R Loizzo, Francesco Menichini, Rosa Tundis
ABSTRACT
The aim of this study was to examine the bioactivity of the methanol fraction (MF) and n-hexane fraction (HF) of Thymus capitatus leaves in relation to their constituents analysed by gas chromatography and gas chromatography-mass spectrometry. The effects of T. capitatus on the growth of pathogenic bacteria associated with respiratory diseases (13 gram-positive and 4 gram-negative) were determined using a microdilution method. The MF was particularly effective on Streptococcus pneumoniae and Moraxella catarrhalis. The antioxidant activity was evaluated by 2,2-diphenyl-1-picrylhydrazyl, 2,2'-azino-bis(3-ethylbenzothiazoline-6-sulphonic acid), ferric-reducing antioxidant power and β-carotene bleaching assays. A strong activity using β-carotene bleaching test was observed with the MF (IC50 of 0.7 μg/mL after 30 min of incubation). In the hypoglycaemic test, a selective α-amylase inhibitory activity was detected with the HF begging the most active (IC50 of 422.5 μg/mL). T. capitatus may represent a source of natural bioactive compounds.
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
3,5-Dinitrosalicylic acid, 98%
Sigma-Aldrich
Potassium sodium tartrate tetrahydrate, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (perchloric acid titration)
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Potassium sodium tartrate tetrahydrate, ACS reagent, 99%
Sigma-Aldrich
Potassium sodium tartrate tetrahydrate, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Potassium sodium tartrate tetrahydrate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium sulfate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium sulfate, BioXtra, ≥99.0%
Sigma-Aldrich
3,5-Dinitrosalicylic acid, used in colorimetric determination of reducing sugars
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Sodium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Linoleic acid, technical, 58-74% (GC)
Sigma-Aldrich
Linoleic acid, ≥98%
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
Potassium sodium tartrate tetrahydrate, puriss., 99.0-101.0% (calc. on H2O free substance), meets analytical specification of Ph. Eur., BP, USP, FC, E 337
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications