Merck
  • Transforming Growth Factor β Signaling in Colorectal Cancer Cells With Microsatellite Instability Despite Biallelic Mutations in TGFBR2.
Transforming Growth Factor β Signaling in Colorectal Cancer Cells With Microsatellite Instability Despite Biallelic Mutations in TGFBR2.
Gastroenterology (2015-03-05)
Noel F C C de Miranda, Maarten van Dinther, Brendy E W M van den Akker, Tom van Wezel, Peter ten Dijke, Hans Morreau
ABSTRACT
Most colorectal cancer (CRC) cells with high levels of microsatellite instability (MSI-H) accumulate mutations at a microsatellite sequence in the gene encoding transforming growth factor β receptor II (TGFBR2). TGFβ signaling therefore is believed to be defective in these tumors, although CRC cells with TGFBR2 mutations have been reported to remain sensitive to TGFβ. We investigated how TGFβ signaling might continue in MSI-H CRC cells. We sequenced the 10-adenines microsatellite sequence in the TGFBR2 gene of 32 MSI-H colon cancer tissues and 6 cell lines (HCT116, LS180, LS411N, RKO, SW48, and SW837). Activation of TGFβ signaling was detected by SMAD2 phosphorylation and through use of a TGFβ-responsive reporter construct in all CRC cell lines. Transcripts of TGFBR2 were knocked-down in CRC cells using short hairpin RNA. Full-length and mutant forms of TGFBR2 were expressed in LS411N cells, which do not respond to TGFβ, and their activities were measured. SMAD2 was phosphorylated in most MSI-H CRC tissues (strong detection in 44% and weak detection in 34% of MSI-H tumors). Phosphorylation of SMAD2 in MSI-H cells required TGFBR2—even the form encoding a frameshift mutation. Transcription and translation of TGFBR2 with a 1-nucleotide deletion at its microsatellite sequence still produced a full-length TGFBR2 protein. However, protein expression required preservation of the TGFBR2 microsatellite sequence; cells in which this sequence was replaced with a synonymous nonmicrosatellite sequence did not produce functional TGFBR2 protein. TGFβ signaling remains active in some MSI-H CRC cells despite the presence of frameshift mutations in the TGFBR2 gene because the mutated gene still expresses a functional protein. Strategies to reactivate TGFβ signaling in colorectal tumors might not be warranted, and the functional effects of mutations at other regions of microsatellite instability should be evaluated.
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Formamide, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Formamide, BioReagent, ≥99.5% (GC), for molecular biology
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Formamide solution, NMR reference standard, 90% in DMSO-d6 (99.9 atom % D), NMR tube size 10 mm × 8 in.
Supelco
Formamide solution, NMR reference standard, 90% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse