Merck
  • Gastrointestinal behavior of nano- and microsized fenofibrate: In vivo evaluation in man and in vitro simulation by assessment of the permeation potential.
Gastrointestinal behavior of nano- and microsized fenofibrate: In vivo evaluation in man and in vitro simulation by assessment of the permeation potential.
European journal of pharmaceutical sciences : official journal of the European Federation for Pharmaceutical Sciences (2015-05-26)
Bart Hens, Joachim Brouwers, Maura Corsetti, Patrick Augustijns
ABSTRACT
The purpose of this study was (i) to evaluate the gastrointestinal behavior of micro- and nanosized fenofibrate in humans and (ii) to develop a simple yet qualitatively predictive in vitro setup that simulates the observed absorption-determining factors. Commercially available micro- and nanoparticles of fenofibrate (Lipanthyl® and Lipanthylnano®, respectively) were administered orally to five healthy volunteers in fasting and postprandial conditions. Intraluminal and systemic drug concentrations were determined as reference data for the development of a predictive in vitro setup. To capture the observed solubility/permeability interplay, in vitro dissolution testing was performed in the presence of a permeation bag with sink conditions. In fasting conditions, intake of nanosized fenofibrate generated increased duodenal concentrations compared to microsized fenofibrate, which was reflected in an improved systemic exposure. In postprandial conditions, duodenal concentrations were greatly enhanced for both formulations, however without an accompanying increase in systemic exposure. It appeared that micellar encapsulation of the highly lipohilic fenofibrate limited its potential to permeate from fed state intestinal fluids. To capture these in vivo observations in an in vitro setup, classic dissolution testing was combined with permeation assessment into a permeation bag with sink conditions. In case of fasting conditions, the dissolution/permeation approach allowed for an improved discriminative power between micro- and nanosized fenofibrate by better simulating the dynamic interplay of dissolution and absorption. In case of postprandial conditions, the observed solubility-permeability interplay could be simulated using the dissolution/permeation approach in combination with biorelevant media (FeSSGFFortimel and FeSSIF-V2) to mimic micellar entrapment and reduced permeation potential of fenofibrate. For the first time, reduced permeation of a lipophilic drug despite increased intraluminal concentrations, was demonstrated in humans. Dissolution testing using biorelevant media in combination with permeation assessment into a sink permeation bag appeared to be a simple yet pragmatic approach to capture this solubility-permeability interplay in early formulation evaluation.
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Fenofibrate, ≥99%, powder
Sigma-Aldrich
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Sodium Acetate Anhydrous, >99%, FG
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Sodium oleate, ≥95% (capillary GC)
Sigma-Aldrich
Sodium oleate, ≥99%
Sigma-Aldrich
Sodium oleate, ≥82% (fatty acids), powder
Sigma-Aldrich
Methanol solution, NMR reference standard, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%