Skip to Content
Merck
All Photos(1)

Key Documents

26125

Sigma-Aldrich

Sodium oleate

≥82% (fatty acids), powder

Synonym(s):

cis-9-Octadecenoic acid sodium salt, Oleic acid sodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
$84.60
15 KG
inquire

$84.60

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 KG
$84.60
15 KG
inquire

About This Item

Linear Formula:
CH3(CH2)7CH=CH(CH2)7COONa
CAS Number:
Molecular Weight:
304.44
Beilstein:
4046357
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

$84.60

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

≥82% (fatty acids)

form

powder

impurities

≤0.5% free alkali (as NaOH)

mp

232-235 °C (lit.)

anion traces

chloride (Cl-): ≤2000 mg/kg

functional group

ester

lipid type

unsaturated FAs

storage temp.

room temp

SMILES string

[Na+].CCCCCCCC\C=C/CCCCCCCC([O-])=O

InChI

1S/C18H34O2.Na/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20;/h9-10H,2-8,11-17H2,1H3,(H,19,20);/q;+1/p-1/b10-9-;

InChI key

BCKXLBQYZLBQEK-KVVVOXFISA-M

Looking for similar products? Visit Product Comparison Guide

Application

Oleic acid is used to investigate effects on apolipoprotein B synthesis, degradation and secretion in cultured cells.

Biochem/physiol Actions

Activates protein kinase C in hepatocytes.
Oleic acid increases hepatic secretion of apolipoprotein B100 in hepatocyte cell lines and in mice . Oleic acid inhibits apolipoprotein B100 secretion at higher physiologic doses .

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna Patrícya Florentino et al.
Frontiers in bioengineering and biotechnology, 8, 550253-550253 (2020-11-17)
This research assessed the microbiological suitability of oleate degradation coupled to sulfidogenesis by enriching communities from anaerobic sludge treating dairy products with S0, SO 3 2 - , SO 4 2 - , and
Tsuguhito Ota et al.
The Journal of clinical investigation, 118(1), 316-332 (2007-12-07)
ER stress can cause hepatic insulin resistance and steatosis. Increased VLDL secretion could protect the liver from ER stress-induced steatosis, but the effect of lipid-induced ER stress on the secretion of VLDL is unknown. To determine the effect of lipids
Simone De Camillis et al.
Nanoscale, 12(39), 20347-20355 (2020-10-03)
Upconversion nanoparticles (UCNPs) exhibit unique optical properties such as photo-emission stability, large anti-Stokes shift, and long excited-state lifetimes, allowing significant advances in a broad range of applications from biomedical sensing to super-resolution microscopy. In recent years, progress on nanoparticle synthesis
Zhongyuan Chen et al.
Theranostics, 9(20), 5784-5796 (2019-09-20)
Background & Aims: The use of antisense oligonucleotide-based nanosystems for the detection and regulation of tumor-related gene expression is thought to be a promising approach for cancer diagnostics and therapies. Herein, we report that a cubic-shaped iron oxide nanoparticle (IONC)
Dylan T King et al.
Langmuir : the ACS journal of surfaces and colloids, 27(18), 11381-11393 (2011-08-16)
The prediction of surfactant phase behavior has applications in a wide range of areas. An accurate modeling of liquid phase behavior can aid our understanding of colloidal process or be used to design phases that respond in a defined way

Questions

  1. What is the main impurities in this chemical?

    1 answer
    1. The impurities of this product are not identified. Historical data suggests other salts and fatty acids (such as palmitic) may be present. The maximum allowable amount of sodium hydroxide is 0.5%.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service