Merck
  • The Demethylase Activity of FTO (Fat Mass and Obesity Associated Protein) Is Required for Preadipocyte Differentiation.
The Demethylase Activity of FTO (Fat Mass and Obesity Associated Protein) Is Required for Preadipocyte Differentiation.
PloS one (2015-07-29)
Meizi Zhang, Ying Zhang, Jun Ma, Feima Guo, Qian Cao, Yu Zhang, Bin Zhou, Jijie Chai, Wenqing Zhao, Renbin Zhao
ABSTRACT
FTO (fat mass and obesity associated gene) was genetically identified to be associated with body mass index (BMI), presumably through functional regulation of energy homeostasis. However, the cellular and molecular mechanisms by which FTO functions remain largely unknown. Using 3T3-L1 preadipocyte as a model to study the role of FTO in adipogenesis, we demonstrated that FTO is functionally required for 3T3-L1 differentiation. FTO knock-down with siRNA inhibited preadipocyte differentiation, whereas ectopic over-expression of FTO enhanced the process. The demethylase activity of FTO is required for differentiation. Level of N6-methyladenosine (m6A) is decreased in cells over-expressing FTO. In contrast, overexpression of R96Q, a FTO missense mutant lack of demethylase activity, had no effect on cellular m6A level and impeded differentiation. Treatment with Rosiglitazone, a PPARγ agonist, could overcome the differentiation inhibition imposed by R96Q mutant, suggesting the effect of FTO is mediated through PPARγ.
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99% (HPLC), powder
Sigma-Aldrich
Oil Red O, certified by the Biological Stain Commission
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99%, BioUltra
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Rosiglitazone, ≥98% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
N,O-Bis(trimethylsilyl)acetamide, synthesis grade, ≥95%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder