Merck
  • MicroRNA-335 and -543 suppress bone metastasis in prostate cancer via targeting endothelial nitric oxide synthase.
MicroRNA-335 and -543 suppress bone metastasis in prostate cancer via targeting endothelial nitric oxide synthase.
International journal of molecular medicine (2015-12-10)
Qizhong Fu, Xianfeng Liu, Ying Liu, Jianxun Yang, Guangyao Lv, Shengfang Dong
ABSTRACT
Skeletal metastasis is the major problem in the management of prostate cancer (PCa). Even though the regulatory role of microRNAs (miRNAs) in the control of tumor metastases has been well described in numerous types of cancer, the importance in bone metastasis of PCa remains largely unknown. In the present study, the differentially expressed miRNAs were identified between the primary PCa and bone metastatic PCa samples by comparing their expression profiling using miRNA microarray, and 4 miRNAs (miR‑335, ‑543, ‑196 and ‑19a) were noted to be significantly downregulated in bone metastasis compared with primary PCa. Among those, the downregulation of 2 miRNAs (miR‑335 and ‑543) was confirmed in a total of 20 paired primary PCa and bone metastasis samples using reverse transcription‑quantitative polymerase chain reaction. Using the online target prediction tool, endothelial nitric oxide synthase (eNOS) was found to be a shared target of miR‑335 and ‑543, which was further verified using the luciferase assay. By examining the expression pattern of eNOS in primary PCa and skeletal metastatic samples, the mRNA and protein expression levels of eNOS were markedly upregulated in the metastatic samples. Furthermore, exogenous overexpression of miR‑335 and ‑543 significantly downregulated the expression level of eNOS, and substantially compromised the ability of migration and invasion in vitro. These findings suggested that miR‑335 and ‑543 are associated with bone metastasis of PCa and indicated that they may have important roles in the bone metastasis, which may also be clinically used as novel biomarkers in discriminating the different stages of human PCa and predicting bone metastasis.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Glycerol solution, puriss., meets analytical specification of Ph. Eur., BP, 84-88%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Glycerol, puriss., anhydrous, 99.0-101.0% (alkalimetric)
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerol, puriss. p.a., ACS reagent, anhydrous, dist., ≥99.5% (GC)
Sigma-Aldrich
Glycerol, BioUltra, for molecular biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder