Merck
  • Functional fingerprinting of human mesenchymal stem cells using high-throughput RNAi screening.
Functional fingerprinting of human mesenchymal stem cells using high-throughput RNAi screening.
Genome medicine (2015-06-30)
Gerrit Erdmann, Michael Suchanek, Patrick Horn, Fabian Graf, Christian Volz, Thomas Horn, Xian Zhang, Wolfgang Wagner, Anthony D Ho, Michael Boutros
ABSTRACT
Mesenchymal stem cells (MSCs) are promising candidates for cellular therapies ranging from tissue repair in regenerative medicine to immunomodulation in graft versus host disease after allogeneic transplantation or in autoimmune diseases. Nonetheless, progress has been hampered by their enormous phenotypic as well as functional heterogeneity and the lack of uniform standards and guidelines for quality control. In this study, we describe a method to perform cellular phenotyping by high-throughput RNA interference in primary human bone marrow MSCs. We have shown that despite heterogeneity of MSC populations, robust functional assays can be established that are suitable for high-throughput and high-content screening. We profiled primary human MSCs against human fibroblasts. Network analysis showed a kinome fingerprint that differs from human primary fibroblasts as well as fibroblast cell lines. In conclusion, this study shows that high-throughput screening in primary human MSCs can be reliably used for kinome fingerprinting.
Sigma-Aldrich
Selenium, pellets, <5 mm particle size, ≥99.999% trace metals basis
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
Selenium, powder, −100 mesh, 99.99% trace metals basis
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
Selenium, powder, −100 mesh, ≥99.5% trace metals basis
Sigma-Aldrich
Selenium, pellets, <5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99%, BioUltra
Sigma-Aldrich
Sodium azide, BioXtra
Sigma-Aldrich
3-Isobutyl-1-methylxanthine, ≥99% (HPLC), powder
Sigma-Aldrich
Sodium azide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Selenium, foil, 25x25mm, thickness 3mm, 99.95%
Selenium, pellets, < 5mm, ≥99.999%
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
SAFC
L-Glutamine
Sigma-Aldrich
Propidium iodide solution