Merck
  • FGF13 regulates proliferation and differentiation of skeletal muscle by down-regulating Spry1.
FGF13 regulates proliferation and differentiation of skeletal muscle by down-regulating Spry1.
Cell proliferation (2015-08-01)
Hongzhao Lu, Xine Shi, Guofang Wu, Jiayu Zhu, Chengchuang Song, Qiangling Zhang, Gongshe Yang
ABSTRACT
Generally, the secretory forms of FGF are known to regulate cell proliferation, differentiation and morphogenesis by binding to the extracellular domain of cell surface receptors. Intracellular FGFs (FGF11-14) are expressed principally in the nervous system. FGF13 is a microtubule-stabilizing protein that regulates neuronal polarization and migration. Previous studies have reported high expression of FGF13 in cultures of single muscle fibres. However, functions of FGF13 in muscle development have not been explored. Real-time RT-PCR was performed to detect expression of FGF13 during C2C12 muscle cell proliferation and differentiation. To further understand the role of FGF13, its effects on proliferation and differentiation were examined by western blot analyses of cells transfected with FGF13 siRNA or FGF13 expression plasmids, or treated with chemical MEK inhibitors. Effects of FGF13 on related signalling pathways in C2C12 cell proliferation and differentiation were determined. FGF13 inhibited C2C12 cell proliferation by up-regulating p27 mRNA level and by down-regulating Cyclin E protein expression, during cell proliferation. Additionally, FGF13 down-regulated Spry1 protein expression, activating the ERK1/2 pathway by phosphorylation and leading to C2C12 cell differentiation inhibition. Consequently, FGF13 seemed to function as a repressor of myoblast differentiation via the ERK1/2 pathway. Although FGF13 inhibited Spry1 regardless of cell proliferation or differentiation, its pathway activation occurred only during the stage of myoblast differentiation. FGF13 inhibited C2C12 cell proliferation and differentiation by down-regulating Spry1. These findings indicate that FGF13 played a negative regulatory role in skeletal muscle development.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Fgf13
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)