Merck
  • Development of a Fully-automated On-line Oxidation Column-switching HPLC System for the Determination of Endogenous Melatonin in Human Clinical Samples.
Development of a Fully-automated On-line Oxidation Column-switching HPLC System for the Determination of Endogenous Melatonin in Human Clinical Samples.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2015-11-13)
Tsubasa Oyama, Rie Nagai, Mayumi Fujimoto, Ryoko Konishi, Masashi Mita, Keiko Uezono, Kiyoshi Zaitsu, Kenji Hamase
ABSTRACT
A fully-automated on-line oxidation column-switching HPLC system has been developed for the determination of endogenous melatonin in human plasma and saliva. This HPLC system consists of four processes. In the first step, melatonin is fractionated using a reversed-phase C4 column (Proteonavi, 75 mm × 1.0 mm i.d.). In the second step, the obtained melatonin fraction is on-line collected, and oxidized to a highly-fluorescent compound, N-[(6-methoxy-4-oxo-1,4-dihydroquinolin-3-yl)methyl]acetamide (6-MOQMA), by mixing with hydrogen peroxide under alkaline conditions. In the third step, the produced 6-MOQMA is concentrated, and the oxidation reagents are removed using an alkaline resistive reversed-phase column, Asahipak ODP (35 mm × 1.0 mm i.d.). In the final fourth step, the 6-MOQMA is determined by a microbore-ODS column packed with ultrafine particles (CAPCELL PAK C18 IF, 250 mm × 1.0 mm i.d.). The limit of detection of melatonin using this system is about 200 amol/injection, and the determination of endogenous melatonin in a small volume of human physiological fluids, such as 100 μL of plasma and 300 μL of saliva, was successfully accomplished.
Product Description
Supelco
Tetrahydrofuran, HPLC grade, ≥99.9%, inhibitor-free
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Methanol, NMR reference standard
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Sodium carbonate, anhydrous, powder or granules, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent (primary standard), 99.95-100.05% dry basis
Sigma-Aldrich
Sodium carbonate, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, E500, anhydrous, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Hydrogen peroxide solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Sodium carbonate, ACS reagent (primary standard), anhydrous, 99.95-100.05% dry basis
Sigma-Aldrich
Sodium carbonate, ACS reagent, anhydrous, ≥99.5%, powder or granules
Sigma-Aldrich
Sodium carbonate, powder, ≥99.5%, ACS reagent
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
Tetrahydrofuran, contains 250 ppm BHT as inhibitor, ACS reagent, ≥99.0%
Sigma-Aldrich
Methanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Tetrahydrofuran, ACS reagent, ≥99.0%, contains 250 ppm BHT as inhibitor
Sigma-Aldrich
Sodium carbonate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Tetrahydrofuran, ReagentPlus®, ≥99.0%, contains 250 ppm BHT as inhibitor
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%