Merck
  • A procedure for efficient non-viral siRNA transfection of primary human monocytes using nucleofection.
A procedure for efficient non-viral siRNA transfection of primary human monocytes using nucleofection.
Journal of immunological methods (2015-04-22)
Olga Scherer, Marten B Maeß, Saskia Lindner, Ulrike Garscha, Christina Weinigel, Silke Rummler, Oliver Werz, Stefan Lorkowski
ABSTRACT
Monocytes are an important constituent of the innate immune system. Therefore, manipulating gene expression of primary human monocytes is a crucial mean to study and characterize the functions of targeted proteins in monocytes. Gene silencing by transfection of cells with small interfering RNA (siRNA) leading to the degradation of the corresponding mRNA and thus to reduced target protein levels is an important tool to investigate gene and protein function of interest. However, non-viral transfection of primary monocytes is challenging because siRNA uptake by these suspended cells is tricky, and the individual cells vary among different donors and do not proliferate. Here, we describe a procedure for efficient non-viral transfection of primary human monocytes isolated from peripheral blood, which maintains cell viability and cell functions, such as responsiveness to stimuli like LPS and IL-10. Nucleofection was used as an electroporation technique that enables efficient introduction of siRNA and silencing of target genes. Using a modification of our previously published protocol for the fast-proliferating THP-1 monocytic cell line, we transfected primary human monocytes with siRNA targeting 5-lipoxygenase (5-LO). In fact, we successfully downregulated 5-LO mRNA resulting in reduced protein levels and enzymatic activity.
Sigma-Aldrich
MISSION® esiRNA, targeting human ALOX5
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Prostaglandin E2, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
7-Aminoactinomycin D, ~97% (HPLC), powder
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Prostaglandin E2, ≥93% (HPLC), synthetic
Sigma-Aldrich
Prostaglandin E2, synthetic, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Alox5
Sigma-Aldrich
tert-Butyl acetoacetate, reagent grade, 98%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O