Merck
  • The chemotherapeutic potential of doxorubicin-loaded PEG-b-PLGA nanopolymersomes in mouse breast cancer model.
The chemotherapeutic potential of doxorubicin-loaded PEG-b-PLGA nanopolymersomes in mouse breast cancer model.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V (2015-07-15)
Mona Alibolandi, Fatemeh Sadeghi, Khalil Abnous, Fatemeh Atyabi, Mohammad Ramezani, Farzin Hadizadeh
ABSTRACT
Vesicles of mPEG-PLGA block copolymer were developed to deliver a therapeutic quantity of doxorubicin (DOX) for breast cancer treatment. The DOX-loaded nanoparticles (NPs) were prepared by the pH-gradient method and then evaluated in terms of morphology, size, DOX encapsulation efficiency and in vitro drug release mechanism. The PEG-PLGA nanopolymersomes were 134±1.2nm spherical NPs with a narrow size distribution (PDI=0.121). DOX was entrapped in mPEG-PLGA nanopolymersomes with an encapsulation efficiency and a loading content of 91.25±4.27% and 7.3±0.34%, respectively. The DOX-loaded nanopolymersomes were found to be stable, demonstrating no significant change in particle size and encapsulation efficiency (EE%) during the 6-month storage period of the lyophilized powder at 4°C. The nanopolymersomes sustained the release of DOX. In cytotoxicity studies of 4T1 cell line samples, free DOX showed a higher cytotoxicity (IC50=1.76μg/mL) than did DOX-loaded nanopolymersomes (15.82μg/mL) in vitro. In order to evaluate the antitumor efficacy and biodistribution of DOX-loaded nanopolymersomes, murine breast tumors were established on the BALB/c mice, and in vivo studies were performed. The obtained results demonstrated that the prepared drug delivery system was highly effective against a murine breast cancer tumor model and successfully accumulated in the tumor site through an enhanced permeation and retention mechanism. In vivo studies also proved that DOX-loaded nanopolymersomes are stable in blood circulation and could be considered a promising and effective DOX delivery system for breast cancer treatment.
Sigma-Aldrich
Glycolic acid solution, high purity, 70 wt. % in H2O
Sigma-Aldrich
Lactic acid, natural, ≥85%
Supelco
Tetrahydrofuran, HPLC grade, ≥99.9%, inhibitor-free
Sigma-Aldrich
Lactic acid, 85%, FCC
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
Glycolic acid, ReagentPlus®, 99%
Sigma-Aldrich
Doxorubicin hydrochloride, suitable for fluorescence, 98.0-102.0% (HPLC)
Sigma-Aldrich
Glycolic acid solution, technical grade, 70 wt. % in H2O
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
Doxorubicin hydrochloride, 98.0-102.0% (HPLC)
Sigma-Aldrich
Glycolic acid, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
Lactic acid, meets USP testing specifications
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide hydrochloride, ≥99.0% (AT)
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Sigma-Aldrich
1,2-Dichloroethane, anhydrous, 99.8%
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide hydrochloride, crystalline
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide hydrochloride, commercial grade, powder
Sigma-Aldrich
DL-Lactic acid, 85 % (w/w), syrup
Sigma-Aldrich
N-(3-Dimethylaminopropyl)-N′-ethylcarbodiimide hydrochloride, BioXtra
Sigma-Aldrich
Lactic acid solution, ACS reagent, ≥85%