Merck
  • The c-MYC-ABCB5 axis plays a pivotal role in 5-fluorouracil resistance in human colon cancer cells.
The c-MYC-ABCB5 axis plays a pivotal role in 5-fluorouracil resistance in human colon cancer cells.
Journal of cellular and molecular medicine (2015-02-18)
Naruji Kugimiya, Arata Nishimoto, Tohru Hosoyama, Koji Ueno, Tadahiko Enoki, Tao-Sheng Li, Kimikazu Hamano
ABSTRACT
c-MYC overexpression is frequently observed in various cancers including colon cancer and regulates many biological activities such as aberrant cell proliferation, apoptosis, genomic instability, immortalization and drug resistance. However, the mechanism by which c-MYC confers drug resistance remains to be fully elucidated. In this study, we found that the c-MYC expression level in primary colorectal cancer tissues correlated with the recurrence rate following 5-fluorouracil (5-FU)-based adjuvant chemotherapy. Supporting this finding, overexpression of exogenous c-MYC increased the survival rate following 5-FU treatment in human colon cancer cells, and knockdown of endogenous c-MYC decreased it. Furthermore, c-MYC knockdown decreased the expression level of ABCB5, which is involved in 5-FU resistance. Using a chromatin immunoprecipitation assay, we found that c-MYC bound to the ABCB5 promoter region. c-MYC inhibitor (10058-F4) treatment inhibited c-MYC binding to the ABCB5 promoter, leading to a decrease in ABCB5 expression level. ABCB5 knockdown decreased the survival rate following 5-FU treatment as expected, and the ABCB5 expression level was increased in 5-FU-resistant human colon cancer cells. Finally, using a human colon cancer xenograft murine model, we found that the combined 5-FU and 10058-F4 treatment significantly decreased tumorigenicity in nude mice compared with 5-FU or 10058-F4 treatment alone. 10058-F4 treatment decreased the ABCB5 expression level in the presence or absence of 5-FU. In contrast, 5-FU treatment alone increased the ABCB5 expression level. Taken together, these results suggest that c-MYC confers resistance to 5-FU through regulating ABCB5 expression in human colon cancer cells.
Sigma-Aldrich
trans-Platinum(II)diammine dichloride
Sigma-Aldrich
Sodium deoxycholate, BioXtra, ≥98.0% (dry matter, NT)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
cis-Diammineplatinum(II) dichloride, crystalline
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)
Sigma-Aldrich
MISSION® esiRNA, targeting human ABCC5
SAFC
Sodium deoxycholate
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Abcb5
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
cis-Diamineplatinum(II) dichloride, ≥99.9% trace metals basis
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Fluorouracil, meets USP testing specifications
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
5-Fluorouracil, ≥99% (HPLC), powder
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
10058-F4, ≥98% (HPLC), solid