Merck
  • TRIM35 Interacts with pyruvate kinase isoform M2 to suppress the Warburg effect and tumorigenicity in hepatocellular carcinoma.
TRIM35 Interacts with pyruvate kinase isoform M2 to suppress the Warburg effect and tumorigenicity in hepatocellular carcinoma.
Oncogene (2014-09-30)
Z Chen, Z Wang, W Guo, Z Zhang, F Zhao, Y Zhao, D Jia, J Ding, H Wang, M Yao, X He
ABSTRACT
Tripartite motif-containing protein 35 (TRIM35) is a member of RBCC family, which has a highly conserved order consisting of a RING domain followed by one or two B-Box domains and then a coiled-coil domain. We previously identified TRIM35 as a novel tumor suppressor in human hepatocellular carcinoma (HCC). However, the molecular mechanism that TRIM35 uses to suppress tumorigenicity is largely unknown. Pyruvate kinase isoform M2 (PKM2) has been demonstrated to have a central role in metabolic reprogramming during cancer progression. Phosphorylation of PKM2 tyrosine residue 105 (Y105) regulates PKM2 to provide a metabolic advantage to tumor cells, thereby promoting tumor growth. In the present work, mass spectrometry analysis demonstrated an interaction between TRIM35 and PKM2. Co-IP experiments confirmed that TRIM35 interacts with PKM2 and that the coiled-coil domain is required for such an interaction. Furthermore, the coiled-coil domain mediates decreases in the Warburg effect and in the cell proliferation of HCC cells. In addition, TRIM35 suppresses the tumorigenicity of HCC cells through the blockade of PKM2 Y105 phosphorylation. Collectively, our data reveal a new function for TRIM35, which is to regulate the Warburg effect and tumorigenicity through interaction with PKM2 in HCC.
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Dihydrorhodamine 123, ≥95%
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))