Merck
  • The usage of micellar extraction for analysis of fluvastatin in water and wastewater samples.
The usage of micellar extraction for analysis of fluvastatin in water and wastewater samples.
Journal of pharmaceutical and biomedical analysis (2014-12-10)
Marta Hryniewicka, Barbara Starczewska
ABSTRACT
This work illustrates the development of new procedures for the isolation and preconcentration of fluvastatin (FLU) from aqueous solutions. Micellar extraction (ME) combined with high performance liquid chromatography (HPLC-UV) has been successfully applied for this purpose. It was found that the analyte created micelle with anionic sodium dodecylsulfate (SDS) and/or with the binary mixture of surfactants nonionic triton X114 (TX114) and cationic tetra-n-butyloammonium bromide (TBAB). The optimal analytical conditions for the proposed extraction procedures (solution pH, concentration of surfactants, centrifugation time and electrolyte type) were ascertained. The calibration curves were recorded. The linearity ranges for FLU, isolated by SDS and the mixture of TX114/TBAB, were 0.21-28.79 μg mL(-1) and 0.21-16.45 μg mL(-1) with limit of detection (LOD) 0.19 μg mL(-1) and 0.14 μg mL(-1), respectively. The recoveries afforded by the proposed methods were high, approximately 97%. These preconcentration procedures were applied for the isolation of the statin from water and wastewater samples taken from the local rivers and wastewater treatment plants.
Sigma-Aldrich
Tetrabutylammonium bromide, ACS reagent, ≥98.0%
Sigma-Aldrich
Fluvastatin sodium hydrate, ≥98% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Supelco
Hexadecyltrimethylammonium bromide, suitable for ion pair chromatography, LiChropur
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioUltra, for molecular biology, ≥99.0% (AT)
Supelco
Hexadecyltrimethylammonium bromide, analytical standard
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Tetrabutylammonium bromide solution, 50 wt. % in H2O
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioXtra, ≥99%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥98%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, for molecular biology, ≥99%
Sigma-Aldrich
Tetrabutylammonium bromide, ReagentPlus®, ≥99.0%
Supelco
Tetrabutylammonium bromide, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, ≥96.0% (AT)
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
1-Octanesulfonic acid sodium salt, ≥98%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
1-Octanesulfonic acid sodium salt, BioXtra