Merck
  • Two antigenotoxic chalcone glycosides from Mentha longifolia subsp. longifolia.
Two antigenotoxic chalcone glycosides from Mentha longifolia subsp. longifolia.
Pharmaceutical biology (2014-11-29)
Zuhal Guvenalp, Hilal Ozbek, Mehmet Karadayi, Medine Gulluce, Ayse Kuruuzum-Uz, Bekir Salih, Omur Demirezer
ABSTRACT
Mentha L. (Labiatae) species (mint) with their flavoring properties have been used in food industries for centuries. Besides they have a great importance in drug development and medicinal applications due to various bioactive compounds of several members of the genus. The aim of this study was to isolate bioactive compounds with antimutagenic potential by bio-guided fractionation and determine their structures by spectroscopic methods. The structural elucidation of the isolated compounds was done based on spectroscopic methods, including MALDI-MS, UV, IR, and 2D NMR experiments, and the bio-guided fractionation process was done by using the Ames/Salmonella test system. Henceforth, solely genotoxic and antigenotoxic potential of the new compounds were also confirmed up to 2 µM/plate by using the same test system. Two new chalcone glycosides: (βR)-β,3,2',6'-tetrahydroxy-4-methoxy-4'-O-rutinosyldihydrochalcone and (βR)-β,4,2',6'-tetrahydroxy-4'-O-rutinosyldihydrochalcone, were isolated from Mentha longifolia (L.) Hudson subsp. longifolia, together with known six flavonoid glycosides and one phenolic acid: apigenin-7-O-glucoside, luteolin-7-O-glucoside, apigenin-7-O-rutinoside, luteolin-7-O-rutinoside, apigenin-7-O-glucuronide, luteolin-7-O-glucuronide, rosmarinic acid. According to the antimutagenicity results, both new test compounds significantly inhibited the mutagenic activity of 9-aminoacridine in a dose-dependent manner at the tested concentrations from 0.8 to 2 µM/plate. (βR)-β,4,2',6'-Tetrahydroxy-4'-O-rutinosyldihydrochalcone showed the maximum inhibition rate as 75.94% at 2 µM/plate concentration. This is the first report that two new chalcone glycosides were isolated from Mentha longifolia subsp. longifolia and their antimutagenic potentials by using mutant bacterial tester strains. In conclusion, the two new chalcone glycosides showed a significant antigenotoxic effect on 9-aminoacridine-induced mutagenesis at tested concentrations.
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Supelco
9-Aminoacridine, matrix substance for MALDI-MS, ≥99.5% (HPLC)
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
L-Histidine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
L-Histidine, suitable for cell culture, meets EP, USP testing specifications, from non-animal source
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Potassium phosphate tribasic, reagent grade, ≥98%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium azide, BioXtra
SAFC
L-Histidine
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%