Merck
  • Profiling the role of deacylation-reacylation in the lymphatic transport of a triglyceride-mimetic prodrug.
Profiling the role of deacylation-reacylation in the lymphatic transport of a triglyceride-mimetic prodrug.
Pharmaceutical research (2014-12-03)
Sifei Han, Luojuan Hu, Tim Quach, Jamie S Simpson, Natalie L Trevaskis, Christopher J H Porter
ABSTRACT
Recent studies have demonstrated the potential for a triglyceride (TG) mimetic prodrug to promote the delivery of mycophenolic acid (MPA) to the lymphatic system. Here, the metabolic pathways that facilitate the lymphatic transport of the TG prodrug (1,3-dipalmitoyl-2-mycophenoloyl glycerol, 2-MPA-TG) were examined to better inform the design of next generation prodrugs. In vitro hydrolysis experiments in simulated intestinal conditions and in vivo rat lymphatic transport experiments were conducted in the presence and absence of orlistat and A922500 (inhibitors of lipolysis and TG re-esterification, respectively), to evaluate the importance of 2-MPA-TG digestion and re-esterification of 2-MPA-MG (the 2-monoglyceride derivative) in promoting lymphatic transport. 2-MPA-TG was rapidly hydrolysed to 2-MPA-MG on incubation with fresh bile and pancreatic fluid (BPF), but not in simulated gastric fluid, heat-inactivated BPF or BPF + orlistat. Orlistat markedly decreased lymphatic transport and systemic exposure of 2-MPA-TG derivatives suggesting that inhibition of pancreatic lipase hindered luminal digestion and absorption of the prodrug. A922500 also significantly decreased lymphatic transport of 2-MPA-TG but redirected MPA to the portal blood, suggesting that hindered re-acylation of 2-MPA-MG resulted in intracellular degradation. Incorporation into TG deacylation-reacylation pathways is a critical determinant of the utility of lymph directed TG-mimetic prodrugs.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Serum Triglyceride Determination Kit, 1 kit sufficient for 250 tests
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Supelco
Hydrogen chloride – ethanol solution, ~1.25 M HCl, for GC derivatization, LiChropur
Sigma-Aldrich
Ammonium acetate solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Ammonium formate, ≥99.995% trace metals basis
Sigma-Aldrich
Ammonium acetate, BioUltra, for molecular biology, ≥99.0%
Sigma-Aldrich
Ammonium formate, BioUltra, ≥99.0% (calc. based on dry substance, NT)
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Mecamylamine hydrochloride
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Supelco
Ammonium formate, eluent additive for LC-MS, LiChropur, ≥99.0%
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Propylene glycol monomethyl ether acetate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Ammonium formate, reagent grade, 97%
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis