Merck
  • Construction and physiochemical characterisation of a multi-composite, potential oral vaccine delivery system (VDS).
Construction and physiochemical characterisation of a multi-composite, potential oral vaccine delivery system (VDS).
International journal of pharmaceutics (2014-04-01)
Marie W Pettit, Paul D R Dyer, John C Mitchell, Peter C Griffiths, Bruce Alexander, Beatrice Cattoz, Richard K Heenan, Stephen M King, Ralf Schweins, Frank Pullen, Stephen R Wicks, Simon C W Richardson
ABSTRACT
An increasing human population requires a secure food supply and a cost effective, oral vaccine delivery system for livestock would help facilitate this end. Recombinant antigen adsorbed onto silica beads and coated with myristic acid, was released (∼15% (w/v)) over 24 h at pH 8.8. At pH 2, the myristic acid acted as an enteric coating, protecting the antigen from a variety of proteases. The antigen adsorbed onto silica particles, coated in myristic acid had a conserved secondary structure (measured by circular dichroism (CD) spectroscopy) following its pH-triggered release. Small angle neutron scattering (SANS) was used to measure the thickness of the adsorbed antigen, finding that its adsorbed conformation was slightly greater than its solution radius of gyration, i.e. 120-160 Å. The addition of myristic acid led to a further increase in particle size, with scattering data consistent with an acid thickness slightly greater than a monolayer of fully extended alkyl chains and a degree of hydration of around 50%. Whilst adsorbed onto the silica and coated in myristic acid, the protein was stable over 14 days at 42 °C, indicating a reduced need for cold chain storage. These data indicate that further investigation is warranted into the development of this technology.
Product Description
Supelco
Sodium hydroxide concentrate, 0.1 M NaOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium bicarbonate, tested according to Ph. Eur.
Supelco
Myristic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Supelco
Myristic acid, analytical standard
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Supelco
Sodium hydroxide solution, 49-51% in water, eluent for IC
Sigma-Aldrich
Sodium azide, purum p.a., ≥99.0% (T)
Sigma-Aldrich
Sodium azide, BioUltra, ≥99.5% (T)
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, for molecular biology, ≥99.0% (GC)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Ammonium acetate solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
Potassium acetate solution, BioUltra, for molecular biology, 5 M in H2O
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Myristic acid, natural, ≥98.5%, FG
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Lithium-7Li chloride, 99 atom % 7Li, 99% (CP)
Supelco
Acrylamide, analytical standard
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Myristic acid, ≥95%, FCC, FG
Sigma-Aldrich
Ammonium acetate, BioUltra, for molecular biology, ≥99.0%