Merck
  • Mechanism of hydroxyl radical generation from biochar suspensions: Implications to diethyl phthalate degradation.
Mechanism of hydroxyl radical generation from biochar suspensions: Implications to diethyl phthalate degradation.
Bioresource technology (2014-12-03)
Guodong Fang, Changyin Zhu, Dionysios D Dionysiou, Juan Gao, Dongmei Zhou
ABSTRACT
This paper investigated hydroxyl radical (OH) generation from biochar suspensions for diethyl phthalate (DEP) degradation in the presence of oxygen. Electron paramagnetic resonance (EPR) coupled with a salicylic acid trapping method were used to detect free radicals in biochar and verify OH generation from biochar suspensions. Free radicals (FRs) in biochar could induce OH generation, and ≈12 spins of FRs were consumed to produce one trapped [OH] molecule. The proposed mechanism of OH generation was that FRs in biochar transferred electrons to O2 to produce the superoxide radical anion and hydrogen peroxide, which reacted further with FRs to produce OH. Free radical-quenching studies utilizing superoxide dismutase, catalase, and deferoxamine as scavengers were used to testify this mechanism. Furthermore, OH generated from biochar suspensions could degrade DEP efficiently. These findings of this study provide new insights into the physicochemical properties and environmental implications of biochar.
Sigma-Aldrich
Pyrocatechol, ≥99%
Sigma-Aldrich
Potassium bromide, BioXtra, ≥99.0%
Sigma-Aldrich
Deferoxamine mesylate salt, powder, ≥92.5% (TLC)
Sigma-Aldrich
Potassium bromide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Pyrocatechol, purified by sublimation, ≥99.5%
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Potassium bromide, anhydrous, powder, 99.95% trace metals basis
Sigma-Aldrich
2,3-Dihydroxybenzoic acid, 99%
Supelco
5,5-Dimethyl-1-pyrroline N-oxide, for ESR-spectroscopy
Deferoxamine mesylate salt, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Potassium bromide, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Potassium bromide, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
Potassium bromide, ≥99% trace metals basis
Sigma-Aldrich
Potassium bromide, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%
Sigma-Aldrich
Potassium bromide, ACS reagent, ≥99.0%
Sigma-Aldrich
Diethyl pyrocarbonate, 96% (NT)
Sigma-Aldrich
2,5-Dihydroxybenzoic acid, 98%
Supelco
2,5-Dihydroxybenzoic acid, >99.0% (HPLC)
Sigma-Aldrich
1,2-Dihydroxybenzene, ReagentPlus®, ≥99%
Sigma-Aldrich
Diethyl pyrocarbonate, 96% (NT)
Sigma-Aldrich
Diethyl pyrocarbonate, 96% (NT)
Supelco
2,5-Dihydroxybenzoic acid, ≥99.5% (HPLC), Ultra pure
Supelco
2,5-Dihydroxybenzoic acid, analytical standard