Merck
  • Oral delivery of highly lipophilic poorly water-soluble drugs: spray-dried dispersions to improve oral absorption and enable high-dose toxicology studies of a P2Y1 antagonist.
Oral delivery of highly lipophilic poorly water-soluble drugs: spray-dried dispersions to improve oral absorption and enable high-dose toxicology studies of a P2Y1 antagonist.
Journal of pharmaceutical sciences (2014-10-14)
Xue-Qing Chen, Kevin Stefanski, Hong Shen, Christine Huang, Christian Caporuscio, Wu Yang, Patrick Lam, Ching Su, Olafur Gudmundsson, Michael Hageman
ABSTRACT
BMS-B is a highly lipophilic compound (clog P 7.72) with poor aqueous solubility (<10 ng/mL at pH 1 and 6.5). The compound exhibits low bioavailability in preclinical species when dosed as cosolvent solution formulations, with reduced exposure upon dose escalation. The purpose of this study was to evaluate spray-dried dispersions (SDDs) for enhancing oral exposure and enabling toxicology studies of BMS-B. SDD solids of BMS-B were prepared with 10%-25% drug in hydroxypropyl methylcellulose acetate succinate and showed an enhanced dissolution profile relative to the neat form of the compound. When dosed in rats and monkeys at 5 mg/kg, the SDD exhibited comparable exposure relative to the solution formulation. The SDD was also dosed in rats at 200 and 400 mg/kg and showed dose-proportional exposure compared to the solution formulation. Based on in vitro and in vivo data, the SDD formulation was selected for the toxicology study of BMS-B in rats. In summary, although the SDD approach could be quite challenging for highly lipophilic compounds because of the limitation on wetting and dissolution, the present study demonstrated that SDD can be applied in drug discovery to enhance oral exposure and enable preclinical toxicology studies of highly lipophilic poorly water-soluble compounds.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Supelco
Alpha Tocopherol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Supelco
Tetrahydrofuran, analytical standard
Supelco
Tetrahydrofuran, Selectophore, ≥99.5%
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Supelco
Tetrahydrofuran, HPLC grade, ≥99.9%, inhibitor-free
Supelco
(±)-α-Tocopherol, analytical standard
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
(±)-α-Tocopherol, synthetic, ≥96% (HPLC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Lysine hydrochloride, European Pharmacopoeia (EP) Reference Standard
USP
Alpha Tocopherol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
(±)-α-Tocopherol, tested according to Ph. Eur.
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Tetrahydrofuran, inhibitor-free, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Tetrahydrofuran, ACS reagent, ≥99.0%, contains 250 ppm BHT as inhibitor
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Supelco
Residual Solvent - Acetonitrile, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Tetrahydrofuran, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Tetrahydrofuran, ReagentPlus®, ≥99.0%, contains 250 ppm BHT as inhibitor