Merck
  • Dual core quantum dots for highly quantitative ratiometric detection of trypsin activity in cystic fibrosis patients.
Dual core quantum dots for highly quantitative ratiometric detection of trypsin activity in cystic fibrosis patients.
Nanoscale (2014-10-03)
Iván Castelló Serrano, Georgiana Stoica, Alba Matas Adams, Emilio Palomares
ABSTRACT
We present herein two colour encoded silica nanospheres (2nanoSi) for the fluorescence quantitative ratiometric determination of trypsin in humans. Current detection methods for cystic fibrosis diagnosis are slow, costly and suffer from false positives. The 2nanoSi proved to be a highly sensitive, fast (minutes), and single-step approach nanosensor for the screening and diagnosis of cystic fibrosis, allowing the quantification of trypsin concentrations in a wide range relevant for clinical applications (25-350 μg L(-1)). Furthermore, as trypsin is directly related to the development of cystic fibrosis (CF), different human genotypes, i.e. CF homozygotic, CF heterozygotic, and unaffected, respectively, can be determined using our 2nanoSi nanospheres. We anticipate the 2nanoSi system to be a starting point for non-invasive, easy-to-use and cost effective ratiometric fluorescent biomarkers for recessive genetic diseases like human cystic fibrosis. In a screening program in which the goal is to detect disease and also the carrier status, early diagnosis could be of great help.
Sigma-Aldrich
Ammonia solution, 4 M in methanol
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Sigma-Aldrich
Ammonia-14N, 99.99 atom % 14N
Sigma-Aldrich
Ammonia solution, 2.0 M in methanol
Sigma-Aldrich
Ammonia solution, 2.0 M in ethanol
Sigma-Aldrich
Ammonia solution, 0.4 M in dioxane
Sigma-Aldrich
Ammonia solution, 2.0 M in isopropanol
Sigma-Aldrich
Ammonia solution, 7 N in methanol
Sigma-Aldrich
Cadmium acetate dihydrate, purum p.a., ≥98.0% (KT)
Sigma-Aldrich
Cadmium acetate dihydrate, reagent grade, 98%
Sigma-Aldrich
(3-Aminopropyl)trimethoxysilane, 97%
Sigma-Aldrich
Trioctylphosphine oxide, technical grade, 90%
Sigma-Aldrich
Cadmium oxide, ≥99.99% trace metals basis
Sigma-Aldrich
Trioctylphosphine oxide, ReagentPlus®, 99%
Supelco
1-Octadecene, analytical standard, ≥99.0% (GC)
Supelco
Toluene, analytical standard
Sigma-Aldrich
1-Octadecene, ≥95.0% (GC)
Sigma-Aldrich
Trioctylphosphine oxide, for extraction analysis, ≥98.5%
Sigma-Aldrich
Cadmium oxide, powder, 99.5% trace metals basis
Sigma-Aldrich
1-Octadecene, technical grade, 90%
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Supelco
Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, suitable for HPLC, 99.9%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluene, Laboratory Reagent, ≥99.3%
Supelco
Residual Solvent - Toluene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Toluene, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Toluene, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.7% (GC)