Skip to Content
Merck
All Photos(1)

Key Documents

718939

Sigma-Aldrich

Ammonia solution

0.4 M in THF

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH3
CAS Number:
Molecular Weight:
17.03
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.21

form

liquid

Quality Level

concentration

0.30-0.50 M (by NaOH, titration)
0.4 M in THF

technique(s)

tissue culture: suitable (plant)

density

0.881 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

N

InChI

1S/H3N/h1H3

InChI key

QGZKDVFQNNGYKY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ammonia solution was used in the detection of ammonia formed by the degradation of ammonium nitrate using quantum cascade laser (QCL) technology.
Use of ammonia as:
  • A phermone in anuran tadpoles
  • A nutrient in soil for plant nutrition

Studies of contributions of archaea and bacteria to the ammonia-oxidizing potential of soils

Interaction of ammonia and xenon with imidazole glycerol phosphate synthase from Thermotoga maritima

Metabolism of ammonia in Aedes aegypti fat body and midgut tissues

Used by ammonia-oxidizing bacteria

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-7.6 °F

Flash Point(C)

-22 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Y B Manteifel et al.
Physiological research, 60 Suppl 1, S185-S191 (2011-07-23)
In laboratory experiments, conspecific excretions and ammonia solutions evoked avoidance reactions in tadpoles of three anuran species, Bufo bufo, Rana temporaria, and Rana arvalis. A differential sensitivity of ammonia chemoreception was determined for two anuran species. For Bufo bufo tadpoles
Christoph Liebold et al.
Protein science : a publication of the Protein Society, 19(9), 1774-1782 (2010-07-29)
The imidazole glycerol phosphate (ImGP) synthase from the hyperthermophilic bacterium Thermotoga maritima is a 1:1 complex of the glutaminase subunit HisH and the cyclase subunit HisF. It has been proposed that ammonia generated by HisH is transported through a channel
I. Arix, et al.,
BMC plant biology, 11 (2011)
Patricia Y Scaraffia et al.
Journal of insect physiology, 56(9), 1040-1049 (2010-03-09)
In order to understand at the tissue level how Aedes aegypti copes with toxic ammonia concentrations that result from the rapid metabolism of blood meal proteins, we investigated the incorporation of (15)N from (15)NH(4)Cl into amino acids using an in
Anne E Taylor et al.
Applied and environmental microbiology, 76(23), 7691-7698 (2010-10-05)
A method was developed to determine the contributions of ammonia-oxidizing archaea (AOA) and ammonia-oxidizing bacteria (AOB) to the nitrification potentials (NPs) of soils taken from forest, pasture, cropped, and fallowed (19 years) lands. Soil slurries were exposed to acetylene to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service