Merck
  • Expression of NEDD9 in hepatocellular carcinoma and its clinical significance.
Expression of NEDD9 in hepatocellular carcinoma and its clinical significance.
Oncology reports (2015-03-31)
Peng Lu, Zhi-Peng Wang, Zheng Dang, Zhi-Gang Zheng, Xiao Li, Liang Zhou, Rui Ding, Shu-Qiang Yue, Ke-Feng Dou
ABSTRACT
Neural precursor cell expressed, developmentally downregulated 9 (NEDD9) plays an integral role in natural and pathological cell biology. Overexpression of NEDD9 protein has been correlated with poor prognosis in various types of cancer. However, few available data address the precise function of the NEDD9 gene in hepatocellular carcinoma (HCC). In the present study, we investigated NEDD9 expression in 40 primary human HCC tissues compared with matched adjacent non-tumor hepatic tissues using RT-qPCR and western blot analysis. Immunohistochemistry was performed to analyze the correlations between NEDD9 expression and clinicopathological factors. Statistical analyses were applied to derive prognostic values of NEDD9 in HCC. The results showed that the NEDD9 mRNA and protein expression levels in HCC tissues were significantly higher than those in matched adjacent non-tumor hepatic tissues. High NEDD9 expression was correlated with larger tumor size, advanced tumor grade, metastasis, intrahepatic venous invasion and high UICC TNM stages in HCC patients. Patients with high NEDD9 expression levels exhibited poorer recurrence-free and overall survival than those with a low NEDD9 expression. Additionally, NEDD9 expression status was an independent prognostic factor for survival. This correlation remained significant in patients with early-stage HCC or with normal serum AFP levels. The results of this study suggest that NEDD9 may be a valuable prognostic biomarker for HCC, including early-stage and AFP-normal patients.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%