Merck
  • Simultaneous determination of various bioactive redox components in Shuang-Huang-Lian preparations using a novel three-channel isocratic elution liquid chromatography with electrochemical detection system.
Simultaneous determination of various bioactive redox components in Shuang-Huang-Lian preparations using a novel three-channel isocratic elution liquid chromatography with electrochemical detection system.
Journal of pharmaceutical and biomedical analysis (2014-03-25)
Liangmian Chen, Hideki Hakamata, Fumiyo Kusu, Zhimin Wang, Huimin Gao, Akira Kotani
ABSTRACT
A novel three-channel isocratic elution high performance liquid chromatography with electrochemical detection (3LC-ECD) system was developed for the simultaneous determination of various bioactive redox components in Shuang-Huang-Lian (SHL) preparations. This 3LC-ECD system consists of three isocratic elution flow ways, two switching valves, four columns, and three electrochemical detectors. Through alternately rotating switching valves to change the elution flow way, six caffeoylquinic acids, four flavonoids, and one phenylethanoid glycoside compound were simultaneously determined within 70 min by the present 3LC-ECD method, in which three compounds of protocatechuic aldehyde, propyl gallate, and kaempferol were used as internal standards. The results demonstrated that the present 3LC-ECD method has achieved desired linearity (r>0.999), precision (relative standard deviation <2.5%), accuracy (recovery, 95.6-103.6%), and high sensitivity (limit of detection, 0.11-0.90 ng/mL). In addition, we successfully determined the contents of these 11 bioactive redox components in 14 batches of SHL oral liquid and 12 batches of SHL lyophilized powder for injection produced by different manufacturers in China.
Product Description
Supelco
Kaempferol, analytical standard
Supelco
Scutellarin, analytical standard
Chlorogenic acid, primary reference standard
Sigma-Aldrich
Chlorogenic acid, ≥95% (titration)
Sigma-Aldrich
Kaempferol, ≥90% (HPLC), powder
Sigma-Aldrich
Kaempferol, ≥97.0% (HPLC)
Chlorogenic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Baicalin, 95%
Sigma-Aldrich
Baicalein, 98%
Supelco
Caffeic acid, ≥99.0% (HPLC)
Sigma-Aldrich
Propyl gallate, ≥98%, FCC
Sigma-Aldrich
Propyl gallate, powder
Sigma-Aldrich
Caffeic acid, ≥98.0% (HPLC)
Sigma-Aldrich
Propyl gallate, antioxidant, ≥98.0% (HPLC)
Sigma-Aldrich
Propyl gallate, ≥98.0% (HPLC)
Supelco
Propyl gallate, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Propyl gallate, United States Pharmacopeia (USP) Reference Standard
Baicalin, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Wogonoside, ≥95% (LC/MS-ELSD)
Sigma-Aldrich
4-O-Caffeoylquinic acid, ≥98.0%
Sigma-Aldrich
Neochlorogenic acid, from Lonicera japonica, ≥98.0% (HPLC)