Skip to Content
Merck
All Photos(1)

Documents

Y0001273

Baicalin

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

5,6-Dihydroxy-4-oxo-2-phenyl-4H-1-benzopyran-7-yl-β-D-glucopyranosiduronic acid, Baicalein 7-O-β-D-glucuronide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H18O11
CAS Number:
Molecular Weight:
446.36
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

baicalin

form

solid

manufacturer/tradename

EDQM

mp

202-205 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O[C@@H]1[C@@H](O)[C@@H](O[C@@H]([C@H]1O)C(O)=O)Oc2cc3OC(=CC(=O)c3c(O)c2O)c4ccccc4

InChI

1S/C21H18O11/c22-9-6-10(8-4-2-1-3-5-8)30-11-7-12(14(23)15(24)13(9)11)31-21-18(27)16(25)17(26)19(32-21)20(28)29/h1-7,16-19,21,23-27H,(H,28,29)/t16-,17-,18+,19-,21+/m0/s1

InChI key

IKIIZLYTISPENI-ZFORQUDYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Baicalin EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Caiyun Huang et al.
The British journal of nutrition, 122(2), 152-161 (2019-04-23)
The present study was carried out to evaluate the effect of dietary supplementation of Scutellaria baicalensis extracts (SBE) on intestinal health in terms of morphology, barrier integrity and immune responses in weaned piglets challenged with Escherichia coli K88. A total
Yongtai Zhang et al.
International journal of pharmaceutics, 479(1), 219-226 (2014-12-30)
This study investigated the transdermal permeability of baicalin, a hydrophobic and readily hydrolyzed drug, delivered by glyceryl monooleate (GMO)-based cubic phase gel (CPG) mediated with Transcotol(®) P (TP, diethylene glycol monoethyl ether). A range of CPGs was produced by varying
N R Srinivas
Xenobiotica; the fate of foreign compounds in biological systems, 40(5), 357-367 (2010-03-17)
Baicalin was extensively researched for utility in a number of therapeutic areas owing to its anti-inflammatory, anti-oxidant, anti-bacterial, and anti-cancer properties. A number of preclinical studies, in vitro work, and mechanistic studies were performed to understand the absorption, distribution, metabolism
Amirhossein Sahebkar
Cutaneous and ocular toxicology, 31(3), 226-234 (2011-11-24)
Sulfur mustard (SM) is a bifunctional alkylating agent with strong blistering, irritant, mutagenic and cytotoxic properties. SM has been widely deployed as a chemical warfare agent for over a century, leading to extensive casualties. Skin is among the first and
Jie Zhen et al.
The American journal of Chinese medicine, 49(1), 147-161 (2020-12-30)
Acute pancreatitis (AP) is a kind of reversible inflammatory process of the exocrine pancreas. During the process, systemic inflammatory syndromes are involved, which relates closely to inflammatory mediators. Baicalin is a type of flavone compound extracted from Scutellaria baicalensis Georgi

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service