Skip to Content
Merck
All Photos(2)

Documents

73577

Supelco

Scutellarin

analytical standard

Synonym(s):

4′,5,6,7-Tetrahydroxyflavone 7-glucuronide, 5,6-Dihydroxy-2-(4-hydroxyphenyl)-4-oxo-4H-1-benzopyran-7-yl-β-D-glucopyranosiduronic acid, 7-(β-D-Glucopyranuronosyloxy)-5,6-dihydroxy-2-(4-hydroxyphenyl)-4H-1-benzopyran-4-one, Breviscapin, Scutellarein-7-O-β-D-glucuronide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H18O12
CAS Number:
Molecular Weight:
462.36
Beilstein:
71779
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

O[C@@H]1[C@@H](O)[C@@H](O[C@@H]([C@H]1O)C(O)=O)Oc2cc3OC(=CC(=O)c3c(O)c2O)c4ccc(O)cc4

InChI

1S/C21H18O12/c22-8-3-1-7(2-4-8)10-5-9(23)13-11(31-10)6-12(14(24)15(13)25)32-21-18(28)16(26)17(27)19(33-21)20(29)30/h1-6,16-19,21-22,24-28H,(H,29,30)/t16-,17-,18+,19-,21+/m0/s1

InChI key

DJSISFGPUUYILV-ZFORQUDYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Scutellarin is flavone glucuronide and an active ingredient of breviscapine. It is widely used for the treatment of cerebral infarction, coronary heart disease and angina pectoris.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Scutellarin may be used as an analytical reference standard for the quantification of the analyte in plasma samples using different chromatography techniques.

Biochem/physiol Actions

Scutellarin is a flavonoid that exhibits a number of cardio- and neuro-protective effects. Scutellarin inhibits p38 activity, ERK phosphorylation, and TGF-β expression and fibrosis in a rat myocardial infarct model. In a cerebral ischemia-reperfusion model, the compound reduced infarct volume and neurological deficits by increasing eNOS expression while inhibiting the induction of iNOS.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shanshan Liu et al.
The Journal of pharmacy and pharmacology, 69(11), 1495-1501 (2017-08-16)
Scutellarin (SCU) is a traditional Chinese medicine used for the treatment of ischaemic cerebrovascular disease, but its clinic applications have been limited due to its poor water solubility, poor bioavailability and short half-life. In comparison with the conventional oral and
Fei Xiong et al.
International journal of pharmaceutics, 421(2), 275-282 (2011-10-18)
Series of monooleate-modified PEG with active carboxylic terminus on the other end (MO-PEG-COOH) were used to modify the lipid emulsions surface to prepare a sterically stabilized lipid emulsions for carrying Traditional Chinese Medicine - breviscapine. Based on the research of
Yue-Qin Zeng et al.
Molecules (Basel, Switzerland), 23(4) (2018-04-13)
Alzheimer's disease (AD) is pathologically characterized by excessive accumulation of amyloid-beta (Aβ) within extracellular spaces of the brain. Aggregation of Aβ has been shown to trigger oxidative stress, inflammation, and neurotoxicity resulting in cognitive dysfunction. In this study, we use
Chunying Gao et al.
Drug metabolism and disposition: the biological fate of chemicals, 40(10), 2009-2020 (2012-07-24)
Scutellarin [scutellarein-7-O-glucuronide (S-7-G)] displayed a unique pharmacokinetic profile in humans after oral administration: the original compound was hardly detected, whereas its isomeric metabolite isoscutellarin [scutellarein-6-O-glucuronide (S-6-G)] had a markedly high exposure. Previous rat study revealed that S-7-G and S-6-G in
Determination of scutellarin in rat plasma by high-performance liquid chromatography with ultraviolet detection
Zhong D, et al.
Journal of Chromatography. B, Biomedical Sciences and Applications, 796(2), 439-444 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service