Merck
  • Defining immunological impact and therapeutic benefit of mild heating in a murine model of arthritis.
Defining immunological impact and therapeutic benefit of mild heating in a murine model of arthritis.
PloS one (2015-03-21)
Chen-Ting Lee, Kathleen M Kokolus, Nicholas D Leigh, Maegan Capitano, Bonnie L Hylander, Elizabeth A Repasky
ABSTRACT
Traditional treatments, including a variety of thermal therapies have been known since ancient times to provide relief from rheumatoid arthritis (RA) symptoms. However, a general absence of information on how heating affects molecular or immunological targets relevant to RA has limited heat treatment (HT) to the category of treatments known as "alternative therapies". In this study, we evaluated the effectiveness of mild HT in a collagen-induced arthritis (CIA) model which has been used in many previous studies to evaluate newer pharmacological approaches for the treatment of RA, and tested whether inflammatory immune activity was altered. We also compared the effect of HT to methotrexate, a well characterized pharmacological treatment for RA. CIA mice were treated with either a single HT for several hours or daily 30 minute HT. Disease progression and macrophage infiltration were evaluated. We found that both HT regimens significantly reduced arthritis disease severity and macrophage infiltration into inflamed joints. Surprisingly, HT was as efficient as methotrexate in controlling disease progression. At the molecular level, HT suppressed TNF-α while increasing production of IL-10. We also observed an induction of HSP70 and a reduction in both NF-κB and HIF-1α in inflamed tissues. Additionally, using activated macrophages in vitro, we found that HT reduced production of pro-inflammatory cytokines, an effect which is correlated to induction of HSF-1 and HSP70 and inhibition of NF-κB and STAT activation. Our findings demonstrate a significant therapeutic benefit of HT in controlling arthritis progression in a clinically relevant mouse model, with an efficacy similar to methotrexate. Mechanistically, HT targets highly relevant anti-inflammatory pathways which strongly support its increased study for use in clinical trials for RA.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Supelco
Potassium hydroxide concentrate, 0.1 M KOH in water (0.1N), Eluent concentrate for IC
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98.0% (NT)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Supelco
3,3′,5,5′-Tetramethylbenzidine, standard for GC
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, ≥98% (TLC)
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidine, tablet, 1 mg substrate per tablet
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Methotrexate for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Potassium hydroxide, anhydrous, ≥99.95% trace metals basis