Merck
  • Molecularly imprinted polymer dedicated to the extraction of glyphosate in natural waters.
Molecularly imprinted polymer dedicated to the extraction of glyphosate in natural waters.
Journal of chromatography. A (2014-08-26)
K Puzio, B Claude, L Amalric, C Berho, E Grellet, S Bayoudh, R Nehmé, Ph Morin
ABSTRACT
Three molecularly imprinted polymers (MIPs) have been synthesized in order to bind efficiently glyphosate (GLY) in natural waters (mineral and underground). Since the target analyte is polar and hydrophilic, electrostatic interactions and hydrogen bonds have been favored with two templates (phenylphosphonic acid and diethyl(α-aminobenzyl)-phosphonic acid) and two functional monomers (1-allyl-2-thiourea and methacrylic acid). MIPs have been assessed by comparison of the recoveries obtained with MIP and NIP (non imprinted polymer) by solid-phase extraction (SPE). The selectivity of MIP versus NIP was satisfactory for the three imprinted polymers with a very straightforward protocol: conditioning of 250 mg of MIP or NIP packed in 3-mL polypropylene cartridges with 3 mL Milli-Q water, loading of Milli-Q water (15 mL) spiked with 5 mg L(-1) of GLY and its metabolite, aminomethylphosphonic acid (AMPA) and elution by 3 mL NH4OH (10mM) or 3 mL HCl (100mM). SPE fractions were directly analyzed by capillary electrophoresis (CE). Thus, the recoveries of both analytes were greater than 80% for all MIPs and less than 25% for most NIPs. Moreover, the MIP prepared with 1-allyl-2-thiourea as functional monomer and phenylphosphonic acid as template displayed a capacity of 0.033 μmol/mg for GLY. However, the substitution of Milli-Q water by mineral water caused the decrease of MIP recoveries, for that, a pretreatment of the sample by ionic exchange resins was set up and succeeded in improving recoveries (about 50% for GLY and 25% for AMPA). Then, groundwaters were spiked with low concentrations of GLY and AMPA (0.5 μgL(-1)) and directly percolated through MIP cartridges. The extractions were carried out by triplicate and the elution fractions were analyzed by UPLC-MS/MS. The results showed no retention of AMPA but a total retention of GLY by MIP.
Product Description
Supelco
L-Histidine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Histidine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phthalic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
N-(Phosphonomethyl)glycine, 96%
SAFC
L-Histidine
Sigma-Aldrich
Ammonium acetate solution, BioUltra, for molecular biology, ~5 M in H2O
Sigma-Aldrich
Ammonium acetate, BioUltra, for molecular biology, ≥99.0%
Sigma-Aldrich
Phthalic acid, reagent grade, 98%
Sigma-Aldrich
Ethylene glycol dimethacrylate, 98%, contains 90-110 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
Phenylphosphonic acid, 98%
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Phthalic acid, puriss. p.a., ≥99.5% (T)
Sigma-Aldrich
N-(Phosphonomethyl)glycine, BioReagent, suitable for plant cell culture
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
L-Histidine, ReagentPlus®, ≥99% (TLC)
Sigma-Aldrich
L-Histidine, suitable for cell culture, meets EP, USP testing specifications, from non-animal source
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Ammonium acetate solution, for molecular biology, 7.5 M
Sigma-Aldrich
Ammonium acetate, for molecular biology, ≥98%
Supelco
Ammonium acetate, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
L-Histidine, BioUltra, ≥99.5% (NT)
Phthalic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%
USP
Cetrimonium bromide, United States Pharmacopeia (USP) Reference Standard
Supelco
Phthalic acid, analytical standard
Sigma-Aldrich
Ammonium acetate, ≥99.99% trace metals basis
Sigma-Aldrich
MES solution, BioUltra, for molecular biology, 0.5 M in H2O
Supelco
Fmoc chloride, for HPLC derivatization, LiChropur, ≥99.0% (HPLC)
Sigma-Aldrich
Fmoc chloride, BioReagent, ≥99.0% (HPLC)
Sigma-Aldrich
Hexadecyltrimethylammonium bromide, BioUltra, for molecular biology, ≥99.0% (AT)