Merck
  • Hypolipidemic and hypoglycemic effects of Centella asiatica (L.) extract in vitro and in vivo.
Hypolipidemic and hypoglycemic effects of Centella asiatica (L.) extract in vitro and in vivo.
Indian journal of experimental biology (2014-10-28)
Nattapon Supkamonseni, Aree Thinkratok, Duangdeun Meksuriyen, Rungrudee Srisawat
ABSTRACT
In vitro study revealed that pancreatic lipase inhibitory activity of C. asiatica extract was significantly higher than rutin but lower than orlistat, an anti-obesity drug. alpha-Amylase inhibitory activities of C. asiatica extract and rutin were significantly lower than acarbose, an anti-diabetic drug. Inhibition of alpha-glucosidase activity by C. asiatica extract, rutin, and acarbose was not different. The in vivo study substantiated the in vitro results. C. asiatica extract (1000 and 2000 mg/4 mL/kg), rutin (1000 mg/4 mL/kg), and orlistat (45 mg/4 mL/kg) significantly decreased plasma glucose, triglyceride and total cholesterol levels in lipid emulsion-induced hyperlipidemic rats at 3 h. However, plasma aspartate aminotransferase and alanine aminotransferase levels did not show significant change. The present work further supports that the C. asiatica extract and its bioactive rutin may help managing hypolipidemic and hypoglycemic effects.
Product Description
Supelco
Sodium carbonate concentrate, 0.1 M Na2CO3 in water, eluent concentrate for IC
Sigma-Aldrich
Acarbose, ≥95% (HPLC)
Sigma-Aldrich
4-Nitrophenyl butyrate, ≥98%
Sigma-Aldrich
Sodium carbonate-12C, 99.9 atom % 12C
Supelco
Acarbose, Pharmaceutical Secondary Standard; Certified Reference Material
Rutoside trihydrate, European Pharmacopoeia (EP) Reference Standard
Acarbose, European Pharmacopoeia (EP) Reference Standard
Supelco
Sodium carbonate concentrate, Na2CO3 72 mM in water, IC eluent concentrate (20x) for Metrosep A Supp 7
Supelco
Sodium carbonate, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
4-Nitrophenyl β-D-glucuronide, ≥99.0% (TLC)
Supelco
Rutin trihydrate, analytical standard
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Sigma-Aldrich
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
4-Nitrophenyl β-D-glucuronide, ≥98% (TLC)
Rutin trihydrate, primary reference standard
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent (primary standard), 99.95-100.05% dry basis
Sigma-Aldrich
Sodium carbonate, ACS reagent (primary standard), anhydrous, 99.95-100.05% dry basis
Sigma-Aldrich
Sodium carbonate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium carbonate, powder, ≥99.5%, ACS reagent
Sigma-Aldrich
Sodium carbonate, ACS reagent, anhydrous, ≥99.5%, powder or granules
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium carbonate, anhydrous, powder or granules, free-flowing, Redi-Dri, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium carbonate, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, E500, anhydrous, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium carbonate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99.5%