Merck
  • Serotonin-induced down-regulation of cell surface serotonin transporter.
Serotonin-induced down-regulation of cell surface serotonin transporter.
Neurochemistry international (2014-01-28)
Trine Nygaard Jørgensen, Peter Møller Christensen, Ulrik Gether
ABSTRACT
The serotonin transporter (SERT) terminates serotonergic signaling and enables refilling of synaptic vesicles by mediating reuptake of serotonin (5-HT) released into the synaptic cleft. The molecular and cellular mechanisms controlling SERT activity and surface expression are not fully understood. Here we demonstrate that the substrate 5-HT itself causes acute down-regulation of SERT cell surface expression. To assess surface SERT expression by ELISA, we used a SERT variant (TacSERT) where the N-terminus of SERT was fused to the intracellular tail of the extracellularly FLAG-tagged single-membrane spanning protein Tac. In stably transfected HEK293 cells, 5-HT caused a dose-dependent reduction in TacSERT surface signal with an EC50 value equivalent to the Km value observed for 5-HT uptake. The 5-HT-induced reduction in surface signal reached maximum within 40-60min and was blocked by the selective SERT inhibitor S-citalopram. 5-HT-induced reduction in SERT expression was further supported by surface biotinylation experiments showing 5-HT-induced reduction in wild type SERT plasma membrane levels. Moreover, preincubation with 5-HT lowered the Vmax for 5-HT uptake in cultured raphe serotonergic neurons, indicting that endogenous cell-surface resident SERT likewise is down-regulated in the presence of substrate.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 20% in H2O
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, for molecular biology, 10% in H2O
Sigma-Aldrich
Serotonin creatinine sulfate monohydrate, powder
Sodium laurilsulfate, European Pharmacopoeia (EP) Reference Standard