Merck
  • Centrifugal partition chromatography elution gradient for isolation of sesquiterpene lactones and flavonoids from Anvillea radiata.
Centrifugal partition chromatography elution gradient for isolation of sesquiterpene lactones and flavonoids from Anvillea radiata.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2015-02-04)
Emilie Destandau, Meryem Alaoui Boukhris, Sandrine Zubrzycki, Mohamed Akssira, Lhoucine El Rhaffari, Claire Elfakir
ABSTRACT
An innovative procedure coupling pressurized solvent extraction and centrifugal partition chromatography (CPC) used in linear gradient elution mode was developed to isolate two pure germacranolides (9α-hydroxyparthenolide and 9β-hydroxyparthenolide) and to separate flavonoids (nepetin, isorhamnetin and jaceosidin) and chlorophyll pigments from aerial parts of Anvillea radiata (Coss.&Durieu). The two main germacranolides recovered using this method represent 2 and 5% of the dried plant material respectively. These molecules were extracted using accelerated solvent extraction with chloroform. After optimization of the CPC method, a two-phase solvent system composed of heptane/ethyl acetate/methanol/water (1:5:1:5 v/v/v/v) was employed in descending mode to isolate the germacranolides. Then the lower phase of a heptane/ethyl acetate/methanol/water (6:5:6:5 v/v/v/v) system was pumped in descending mode to generate a linear elution gradient, progressively decreasing the mobile phase polarity, that enabled the flavonoid compounds to be separated in the same run. The efficiency of the preparative separation was controlled through RP-HPLC analysis of the obtained fractions using UV, evaporative light scattering and mass spectrometry detection. The structural identification of the two germacranolides purified over 99% was established by (1)H NMR and (13)C NMR. The least abundant flavonoids were identified by mass spectrometry.
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, for molecular biology, ≥99.0% (GC)
Supelco
Methanol, analytical standard
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Supelco
Chloroform, analytical standard
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
Heptane, biotech. grade, ≥99%
Sigma-Aldrich
Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Supelco
Tetrahydrofuran, HPLC grade, ≥99.9%, inhibitor-free
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
Heptane, anhydrous, 99%
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, for molecular biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Supelco
Heptane, analytical standard
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Supelco
Tetrahydrofuran, Selectophore, ≥99.5%
Supelco
Tetrahydrofuran, analytical standard
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Methanol, NMR reference standard
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, for molecular biology, ≥99.0% (GC)