Merck
  • Evaluation of 18F-fluoride PET/MR and PET/CT in patients with foot pain of unclear cause.
Evaluation of 18F-fluoride PET/MR and PET/CT in patients with foot pain of unclear cause.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine (2015-02-14)
Isabel Rauscher, Ambros J Beer, Christoph Schaeffeler, Michael Souvatzoglou, Moritz Crönlein, Chlodwig Kirchhoff, Gunther Sandmann, Sebastian Fürst, Robert Kilger, Michael Herz, Sybille Ziegler, Markus Schwaiger, Matthias Eiber
ABSTRACT
Our objective was to compare the quality and diagnostic performance of (18)F-fluoride PET/MR imaging with that of (18)F-fluoride PET/CT imaging in patients with foot pain of unclear cause. Twenty-two patients (9 men, 13 women; mean age, 48 ± 18 y; range, 20-78 y) were prospectively included in this study and underwent a single-injection dual-imaging protocol with (18)F-fluoride PET/CT and PET/MR. At a minimum, the PET/MR protocol included T1-weighted spin echo and proton-density fat-saturated sequences in 2 planes each with simultaneous acquisition of PET over 20 min. PET/CT included a native isotropic (0.6 mm) diagnostic CT scan (80 kV, 165 mAs) and a subsequent PET scan (2 min per bed position). By consensus, 2 masked interpreters randomly assessed both PET datasets for image quality (3-point scale) and for the presence of focal lesions with increased (18)F-fluoride uptake (maximum of 4 lesions). For each dataset (PET/CT vs. PET/MR), the diagnoses were defined using both PET and a morphologic dataset. Standardized uptake values (SUVs) from the 2 devices were compared using linear correlation and Bland-Altman plots. Moreover, we estimated the potential for dose reduction for PET/MR compared with PET/CT considering the longer acquisition time of PET/MR analyzing count rate statistics. Image quality was rated diagnostic for both PET datasets. However, with a mean rating of 3.0/3 for PET/MR and 2.3/3 for PET/CT, image quality was significantly superior for PET/MR (P < 0.0001). The sensitivity of the PET datasets in PET/MR and PET/CT was equivalent, with the same 42 lesions showing focal (18)F-fluoride uptake. In PET/MR, the mean SUVmean was 10.4 (range, 2.0-67.7) and the mean SUVmax was 15.6 (range, 2.9-94.1). In PET/CT, the corresponding mean SUVmean of PET/CT was 10.2 (range, 1.8-55.6) and the mean SUVmax was 16.3 (range, 2.5-117.5), resulting in a high linear correlation coefficient (r = 0.96, P < 0.0001, for SUVmean and for SUVmax). A final consensus interpretation revealed the most frequent main diagnoses to be osteoarthritis, stress fracture, and bone marrow edema. PET/CT was more precise in visualizing osteoarthritis, whereas PET/MR was more specific in nondegenerative pathologies because of the higher soft-tissue and bone marrow contrast. The longer acquisition time of MR compared with CT would potentially allow (18)F-fluoride dose reduction using hybrid (18)F-fluoride PET/MR imaging of at least 50% according to the counting rate analysis. In patients with foot pain of unclear cause, (18)F-fluoride PET/MR is technically feasible and is more robust in terms of image quality and SUV quantification than (18)F-fluoride PET/CT. In most patients, (18)F-fluoride PET/MR provided more diagnostic information at a higher diagnostic certainty than did PET/CT. Thus, PET/MR combines the high sensitivity of (18)F-fluoride PET to pinpoint areas with the dominant disease activity and the specificity of MR imaging for the final diagnosis with the potential for a substantial dose reduction compared with PET/CT.
Sigma-Aldrich
Tantalum(V) ethoxide, 99.98% trace metals basis
Sigma-Aldrich
Gadolinium, chips
Sigma-Aldrich
Gadolinium, −40 mesh, 99% trace rare earth metals basis
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Gadolinium, rod, 50mm, diameter 6.35mm, cast, 99%
Gadolinium, wire reel, 50mm, diameter 0.5mm, hard, 99.9%
Gadolinium, rod, 100mm, diameter 12.5mm, cast, 99%
Gadolinium, rod, 48mm, diameter 3.0mm, cast, 99%
Gadolinium, rod, 50mm, diameter 12.5mm, cast, 99%
Gadolinium, wire reel, 100mm, diameter 0.5mm, hard, 99.9%
Gadolinium, wire reel, 25mm, diameter 0.5mm, hard, 99.9%
Gadolinium, rod, 100mm, diameter 3.0mm, cast, 99%
Gadolinium, rod, 100mm, diameter 6.35mm, cast, 99%
Tantalum(V) ethoxide, packaged for use in deposition systems