Skip to Content
Merck
All Photos(2)

Key Documents

GF98041989

Gadolinium

rod, 50mm, diameter 12.5mm, cast, 99%

Synonym(s):

Gadolinium, GD007925

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$1,780.00

$1,780.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

Select a Size

Change View
1 EA
$1,780.00

About This Item

Linear Formula:
Gd
CAS Number:
Molecular Weight:
157.25
MDL number:
UNSPSC Code:
12141605
PubChem Substance ID:
NACRES:
NA.23

$1,780.00

PRICE WITHOUT NATIONAL TAXES

Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Request a Bulk Order

Assay

≥99%

form

rod

manufacturer/tradename

Goodfellow 980-419-89

resistivity

126 μΩ-cm, 20°C

bp

3273 °C (lit.)

mp

1313 °C (lit.)

density

7.886 g/mL at 25 °C (lit.)

SMILES string

[Gd]

InChI

1S/Gd

InChI key

UIWYJDYFSGRHKR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Water-react 3

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J T Heverhagen et al.
RoFo : Fortschritte auf dem Gebiete der Rontgenstrahlen und der Nuklearmedizin, 186(7), 661-669 (2014-01-31)
Nephrogenic systemic fibrosis (NSF) is a serious, sometimes fatal disease. Findings in recent years have shown that a causal association between gadolinium containing contrast media and NSF is most likely. Therefore, the regulatory authorities have issued guidelines on the use
Anders Bruun Mathiasen et al.
European heart journal, 36(27), 1744-1753 (2015-05-01)
Regenerative treatment with mesenchymal stromal cells (MSCs) has been promising in patients with ischaemic heart failure but needs confirmation in larger randomized trials. We aimed to study effects of intra-myocardial autologous bone marrow-derived MSC treatment in patients with severe ischaemic
Stephanie Nougaret et al.
Radiology, 276(3), 797-808 (2015-05-01)
To investigate magnetic resonance (MR) volumetry of endometrial tumors and its association with deep myometrial invasion, tumor grade, and lymphovascular invasion and to assess the value of apparent diffusion coefficient (ADC) histographic analysis of the whole tumor volume for prediction
John J Green et al.
JACC. Cardiovascular imaging, 5(4), 370-377 (2012-04-14)
The objective of this study was to perform a systematic review and meta-analysis of the predictive value of late gadolinium enhancement (LGE) cardiac magnetic resonance (CMR) for future cardiovascular events and death in hypertrophic cardiomyopathy (HCM). The utility of LGE
Henrik S Thomsen et al.
European radiology, 23(2), 307-318 (2012-08-07)
To update the guidelines of the Contrast Media Safety Committee (CMSC) of the European Society of Urogenital Radiology (ESUR) on nephrogenic systemic fibrosis and gadolinium-based contrast media. Topics reviewed include the history, clinical features and prevalence of nephrogenic systemic fibrosis

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service